Assay Details
| Gene Symbol: MLEC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001303627.1 | NP_001290556.1 | 2 - 3 | 305 | 62 | |
| NM_014730.3 | NP_055545.1 | 2 - 3 | 588 | 62 | ||
| XM_011539031.1 | XP_011537333.1 | 2 - 3 | 5003 | 62 | ||
| XM_011539032.1 | XP_011537334.1 | 3 - 4 | 4691 | 62 | ||
| GenBank mRNA | BC000371.2 | - | 2 - 3 | 537 | 2819615 | 62 |
| BC016297.2 | - | 2 - 3 | 539 | 3832682 | 62 | |
| BI458950.1 | - | 2 - 3 | 307 | 62 | ||
| BU845446.1 | - | 566 | 62 | |||
| D63486.1 | - | 2 - 3 | 543 | 62 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000949.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12012320.hg.1 | ENST00000228506.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012320.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012320.hg.1 | TC12000949.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000412616.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000412616.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000412616.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000412616.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000412616.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000412616.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000412616.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000412616.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000412616.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000412616.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000535413.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000535413.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000535656.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000535656.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000535656.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000535656.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000535656.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000535656.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000545525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000545525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000545525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000545525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000545525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | ENST00000545525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200167583.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200167583.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200167583.hg.1 | TC1200009137.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000412616.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000412616.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000412616.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000412616.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000412616.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000412616.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000412616.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000412616.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000412616.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000412616.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000535413.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000535413.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000535656.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000535656.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000535656.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000535656.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000535656.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000535656.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000545525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000545525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000545525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000545525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000545525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | ENST00000545525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_001303628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009137.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200167587.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200167587.hg.1 | ENST00000535656.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200167587.hg.1 | ENST00000545525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200167587.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200167587.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200167587.hg.1 | ENST00000228506.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200167587.hg.1 | ENST00000535656.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200167587.hg.1 | ENST00000545525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200167587.hg.1 | NM_001303627.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200167587.hg.1 | NM_014730.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200167587.hg.1 | TC1200009137.hg.1 | Clariom D TC |

MLEC
Genomic Map