Assay Details
| Gene Symbol: ZC3H10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032786.2 | NP_116175.1 | 1 - 2 | 61 | ||
| GenBank mRNA | BC018708.1 | - | 1 - 2 | 4111500 | 61 | |
| DA050357.1 | - | 1 - 2 | 105 | 63 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000503.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC018708.1,NM_032786.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12006503.hg.1 | ENST00000257940.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12006503.hg.1 | ENST00000549973.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12006503.hg.1 | NM_032786.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12006503.hg.1 | TC12000503.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000257940.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000257940.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000257940.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000257940.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000257940.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000257940.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000257940.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000257940.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000257940.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000257940.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000257940.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000546903.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000546903.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000546903.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000546903.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000546903.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000546903.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000546903.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000549973.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000549973.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000549973.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000551880.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000551880.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000551880.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000551880.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000551880.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000551880.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000551880.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000551880.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000552345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000552345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000552345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000552345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000552345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000552345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | NM_001303124.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | NM_001303124.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | NM_001303124.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | NM_001303124.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | NM_001303124.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | NM_001303124.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | NM_001303124.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | NM_001303124.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | NM_001303124.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | NM_001303124.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | NM_001303125.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | NM_001303125.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | NM_001303125.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | NM_001303125.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | NM_001303125.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | NM_001303125.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | NM_001303125.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | NM_001303125.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | NM_032786.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | NM_032786.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | NM_032786.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | NM_032786.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | NM_032786.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | NM_032786.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | NM_032786.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | NM_032786.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | NM_032786.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12006504.hg.1 | ENST00000257940.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12006504.hg.1 | ENST00000549973.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12006504.hg.1 | NM_032786.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12006504.hg.1 | TC12000503.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000257940.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000257940.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000257940.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000257940.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000257940.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000257940.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000257940.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000257940.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000257940.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000257940.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000257940.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000546903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000546903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000546903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000546903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000546903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000546903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000546903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000549973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000549973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000549973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000551880.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000551880.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000551880.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000551880.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000551880.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000551880.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000551880.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000551880.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000552345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000552345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000552345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000552345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000552345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000552345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | NM_001303124.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | NM_001303124.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | NM_001303124.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | NM_001303124.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | NM_001303124.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | NM_001303124.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | NM_001303124.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | NM_001303124.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | NM_001303124.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | NM_001303124.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | NM_001303125.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | NM_001303125.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | NM_001303125.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | NM_001303125.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | NM_001303125.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | NM_001303125.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | NM_001303125.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | NM_001303125.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | NM_032786.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | NM_032786.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | NM_032786.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | NM_032786.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | NM_032786.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | NM_032786.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | NM_032786.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | NM_032786.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | NM_032786.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200200467.hg.1 | ENST00000257940.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200200467.hg.1 | ENST00000546903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200200467.hg.1 | ENST00000549973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200200467.hg.1 | ENST00000551880.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200200467.hg.1 | ENST00000552345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200200467.hg.1 | NM_001303124.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200200467.hg.1 | NM_001303125.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200200467.hg.1 | NM_032786.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200200467.hg.1 | ENST00000257940.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200200467.hg.1 | ENST00000546903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200200467.hg.1 | ENST00000549973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200200467.hg.1 | ENST00000551880.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200200467.hg.1 | ENST00000552345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200200467.hg.1 | NM_001303124.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200200467.hg.1 | NM_001303125.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200200467.hg.1 | NM_032786.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200200467.hg.1 | TC1200012645.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200200458.hg.1 | ENST00000257940.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200200458.hg.1 | NM_001303124.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200200458.hg.1 | NM_032786.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200200458.hg.1 | ENST00000257940.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200200458.hg.1 | NM_001303124.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200200458.hg.1 | NM_032786.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200200458.hg.1 | TC1200012645.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200200461.hg.1 | ENST00000257940.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200200461.hg.1 | ENST00000549973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200200461.hg.1 | ENST00000552345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200200461.hg.1 | NM_001303124.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200200461.hg.1 | NM_032786.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200200461.hg.1 | ENST00000257940.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200200461.hg.1 | ENST00000549973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200200461.hg.1 | ENST00000552345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200200461.hg.1 | NM_001303124.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200200461.hg.1 | NM_032786.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200200461.hg.1 | TC1200012645.hg.1 | Clariom D TC |

ZC3H10
Genomic Map