Assay Details
| Gene Symbol: OVCH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198185.4 | NP_937828.3 | 5 - 6 | 559 | 95 | |
| XM_017017659.1 | XP_016873148.1 | 5 - 6 | 1059 | 95 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11001373.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100013141.hg.1 | ENST00000533663.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013141.hg.1 | ENST00000533663.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013141.hg.1 | ENST00000533663.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013141.hg.1 | ENST00000533663.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013141.hg.1 | ENST00000533663.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013141.hg.1 | ENST00000533663.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013141.hg.1 | ENST00000533663.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013141.hg.1 | ENST00000533663.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013141.hg.1 | ENST00000533663.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013141.hg.1 | ENST00000533663.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013141.hg.1 | ENST00000533663.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013141.hg.1 | ENST00000533663.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013141.hg.1 | ENST00000533663.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013141.hg.1 | ENST00000533663.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013141.hg.1 | ENST00000533663.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013141.hg.1 | ENST00000533663.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013141.hg.1 | ENST00000533663.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013141.hg.1 | ENST00000533663.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013141.hg.1 | ENST00000534817.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013141.hg.1 | ENST00000534817.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013141.hg.1 | ENST00000534817.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013141.hg.1 | ENST00000612000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013141.hg.1 | ENST00000612000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013141.hg.1 | ENST00000612000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013141.hg.1 | ENST00000612000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013141.hg.1 | ENST00000612000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013141.hg.1 | ENST00000612000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013141.hg.1 | ENST00000612000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013141.hg.1 | ENST00000612000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013141.hg.1 | ENST00000612000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013141.hg.1 | ENST00000612000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013141.hg.1 | ENST00000612000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013141.hg.1 | ENST00000612000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013141.hg.1 | ENST00000612000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013141.hg.1 | ENST00000612000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013141.hg.1 | ENST00000612000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013141.hg.1 | ENST00000612000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013141.hg.1 | NM_198185.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013141.hg.1 | NM_198185.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013141.hg.1 | NM_198185.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013141.hg.1 | NM_198185.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013141.hg.1 | NM_198185.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013141.hg.1 | NM_198185.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013141.hg.1 | NM_198185.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013141.hg.1 | NM_198185.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013141.hg.1 | NM_198185.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013141.hg.1 | NM_198185.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013141.hg.1 | NM_198185.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013141.hg.1 | NM_198185.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013141.hg.1 | NM_198185.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013141.hg.1 | NM_198185.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013141.hg.1 | NM_198185.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013141.hg.1 | NM_198185.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11017310.hg.1 | ENST00000454689.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11017310.hg.1 | NM_198185.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11017310.hg.1 | TC11001373.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1100203499.hg.1 | ENST00000533663.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203499.hg.1 | ENST00000612000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203499.hg.1 | NM_198185.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100203499.hg.1 | ENST00000533663.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203499.hg.1 | ENST00000612000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203499.hg.1 | NM_198185.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100203499.hg.1 | TC1100013141.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100013141.hg.1 | ENST00000533663.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013141.hg.1 | ENST00000533663.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013141.hg.1 | ENST00000533663.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013141.hg.1 | ENST00000533663.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013141.hg.1 | ENST00000533663.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013141.hg.1 | ENST00000533663.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013141.hg.1 | ENST00000533663.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013141.hg.1 | ENST00000533663.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013141.hg.1 | ENST00000533663.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013141.hg.1 | ENST00000533663.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013141.hg.1 | ENST00000533663.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013141.hg.1 | ENST00000533663.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013141.hg.1 | ENST00000533663.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013141.hg.1 | ENST00000533663.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013141.hg.1 | ENST00000533663.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013141.hg.1 | ENST00000533663.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013141.hg.1 | ENST00000533663.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013141.hg.1 | ENST00000533663.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013141.hg.1 | ENST00000534817.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013141.hg.1 | ENST00000534817.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013141.hg.1 | ENST00000534817.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013141.hg.1 | ENST00000612000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013141.hg.1 | ENST00000612000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013141.hg.1 | ENST00000612000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013141.hg.1 | ENST00000612000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013141.hg.1 | ENST00000612000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013141.hg.1 | ENST00000612000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013141.hg.1 | ENST00000612000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013141.hg.1 | ENST00000612000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013141.hg.1 | ENST00000612000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013141.hg.1 | ENST00000612000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013141.hg.1 | ENST00000612000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013141.hg.1 | ENST00000612000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013141.hg.1 | ENST00000612000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013141.hg.1 | ENST00000612000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013141.hg.1 | ENST00000612000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013141.hg.1 | ENST00000612000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013141.hg.1 | NM_198185.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013141.hg.1 | NM_198185.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013141.hg.1 | NM_198185.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013141.hg.1 | NM_198185.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013141.hg.1 | NM_198185.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013141.hg.1 | NM_198185.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013141.hg.1 | NM_198185.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013141.hg.1 | NM_198185.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013141.hg.1 | NM_198185.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013141.hg.1 | NM_198185.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013141.hg.1 | NM_198185.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013141.hg.1 | NM_198185.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013141.hg.1 | NM_198185.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013141.hg.1 | NM_198185.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013141.hg.1 | NM_198185.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013141.hg.1 | NM_198185.4 | Refseq |

OVCH2
Genomic Map