Assay Details
| Gene Symbol: AFDN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040000.2 | NP_001035089.1 | 9 - 10 | 1365 | 127 | |
| NM_001291964.1 | NP_001278893.1 | 9 - 10 | 1419 | 127 | ||
| XM_005266996.3 | XP_005267053.1 | 9 - 10 | 1484 | 127 | ||
| XM_005266997.2 | XP_005267054.1 | 9 - 10 | 1482 | 127 | ||
| XM_006715491.2 | XP_006715554.1 | 9 - 10 | 1482 | 127 | ||
| XM_011535842.2 | XP_011534144.1 | 9 - 10 | 1484 | 127 | ||
| XM_017010878.1 | XP_016866367.1 | 9 - 10 | 1482 | 127 | ||
| XM_017010879.1 | XP_016866368.1 | 9 - 10 | 1484 | 127 | ||
| XM_017010880.1 | XP_016866369.1 | 9 - 10 | 1463 | 127 | ||
| XM_017010881.1 | XP_016866370.1 | 9 - 10 | 1464 | 127 | ||
| XM_017010882.1 | XP_016866371.1 | 9 - 10 | 1482 | 127 | ||
| XM_017010886.1 | XP_016866375.1 | 9 - 10 | 1482 | 127 | ||
| XM_017010887.1 | XP_016866376.1 | 6 - 7 | 721 | 127 | ||
| GenBank mRNA | AB209420.1 | - | 9 - 10 | 1188 | 127 | |
| AB621809.1 | - | 9 - 10 | 1419 | 127 | ||
| AL161973.1 | - | 2 - 3 | 221 | 127 | ||
More Information
Set Membership: |
|

AFDN
Genomic Map