Assay Details
Gene Symbol: RNF165 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256758.1 | NP_001243687.1 | 4 - 5 | 445 | 80 | |
NM_152470.2 | NP_689683.2 | 6 - 7 | 869 | 80 | ||
GenBank mRNA | AK122819.1 | - | 4 - 5 | 445 | 80 | |
AK127467.1 | - | 6 - 7 | 869 | 80 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC18000168.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
protein polyubiquitination
motor neuron axon guidance
multicellular organism aging
positive regulation of BMP signaling pathway
forelimb morphogenesis
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
proteasome-mediated ubiquitin-dependent protein catabolic process
innervation
muscle structure development
zinc ion binding
ubiquitin protein ligase activity
protein polyubiquitination
motor neuron axon guidance
multicellular organism aging
positive regulation of BMP signaling pathway
forelimb morphogenesis
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
proteasome-mediated ubiquitin-dependent protein catabolic process
innervation
muscle structure development
zinc ion binding
ubiquitin protein ligase activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000269439.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000269439.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000269439.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000269439.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000269439.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000269439.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000269439.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000269439.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000269439.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000269439.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000269439.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000269439.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000269439.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000543885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000543885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000543885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000543885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000543885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000543885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000543885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000543885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000543885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000543885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000586604.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000586604.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000586604.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000586604.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000586604.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000586604.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000586604.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000586604.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000586604.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000586604.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000586604.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000587853.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000587853.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000587853.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000587853.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000587853.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000587853.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000587853.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000588679.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000588679.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000588679.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000588679.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000588679.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000588679.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000588679.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000590330.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000590330.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000590330.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000590330.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000590330.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000592777.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000592777.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000592777.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000592777.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000592777.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000592777.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000592777.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000593230.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000593230.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000593230.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000593230.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000593230.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000593230.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | NM_001256758.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | NM_001256758.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | NM_001256758.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | NM_001256758.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | NM_001256758.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | NM_001256758.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | NM_001256758.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | NM_001256758.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | NM_001256758.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | NM_001256758.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | NM_152470.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | NM_152470.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | NM_152470.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | NM_152470.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | NM_152470.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | NM_152470.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | NM_152470.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | NM_152470.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | NM_152470.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | NM_152470.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | NM_152470.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | NM_152470.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | NR_046357.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | NR_046357.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | NR_046357.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | NR_046357.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | NR_046357.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18001975.hg.1 | ENST00000269439.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18001975.hg.1 | ENST00000543885.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18001975.hg.1 | NM_152470.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18001975.hg.1 | TC18000168.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1800149568.hg.1 | ENST00000269439.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800149568.hg.1 | ENST00000543885.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800149568.hg.1 | ENST00000586604.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800149568.hg.1 | ENST00000592777.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800149568.hg.1 | NM_001256758.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800149568.hg.1 | NM_152470.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800149568.hg.1 | TC1800007220.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800149568.hg.1 | ENST00000269439.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800149568.hg.1 | ENST00000543885.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800149568.hg.1 | ENST00000586604.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800149568.hg.1 | ENST00000592777.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800149568.hg.1 | NM_001256758.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800149568.hg.1 | NM_152470.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000269439.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000269439.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000269439.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000269439.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000269439.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000269439.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000269439.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000269439.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000269439.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000269439.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000269439.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000269439.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000269439.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000543885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000543885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000543885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000543885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000543885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000543885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000543885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000543885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000543885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000543885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000586604.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000586604.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000586604.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000586604.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000586604.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000586604.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000586604.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000586604.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000586604.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000586604.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000586604.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000587853.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000587853.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000587853.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000587853.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000587853.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000587853.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000587853.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000588679.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000588679.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000588679.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000588679.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000588679.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000588679.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000588679.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000590330.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000590330.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000590330.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000590330.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000590330.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000592777.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000592777.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000592777.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000592777.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000592777.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000592777.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000592777.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000593230.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000593230.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000593230.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000593230.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000593230.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000593230.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | NM_001256758.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | NM_001256758.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | NM_001256758.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | NM_001256758.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | NM_001256758.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | NM_001256758.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | NM_001256758.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | NM_001256758.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | NM_001256758.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | NM_001256758.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | NM_152470.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | NM_152470.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | NM_152470.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | NM_152470.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | NM_152470.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | NM_152470.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | NM_152470.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | NM_152470.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | NM_152470.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | NM_152470.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | NM_152470.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | NM_152470.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | NR_046357.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | NR_046357.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | NR_046357.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | NR_046357.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | NR_046357.1 | Refseq |
