Assay Details
| Gene Symbol: TONSL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013432.4 | NP_038460.4 | 2 - 3 | 142 | 115 | |
| XM_011517050.2 | XP_011515352.1 | 2 - 3 | 201 | 115 | ||
| GenBank mRNA | BC008782.2 | - | 1 - 2 | 114 | 3628374 | 115 |
| CN366457.1 | - | 2 - 3 | 166 | 115 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08001751.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC008782.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08023289.hg.1 | TC08001751.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08023289.hg.1 | ENST00000409379.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08023289.hg.1 | ENST00000422691.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08023289.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800183819.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800183819.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800183819.hg.1 | ENST00000409379.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800183819.hg.1 | NM_013432.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800183819.hg.1 | TC0800012214.hg.1 | Clariom D TC |

TONSL
Genomic Map