Assay Details
| Gene Symbol: CACNA2D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000722.3 | NP_000713.2 | 23 - 24 | 2236 | 123 | |
| XM_005250572.2 | XP_005250629.1 | 23 - 24 | 2328 | 123 | ||
| XM_006716118.2 | XP_006716181.1 | 24 - 25 | 2402 | 123 | ||
| XM_006716119.3 | XP_006716182.1 | 24 - 25 | 4185 | 123 | ||
| XM_006716120.3 | XP_006716183.1 | 24 - 25 | 3627 | 123 | ||
| XM_006716121.3 | XP_006716184.1 | 6 - 7 | 1251 | 123 | ||
| XM_011516570.2 | XP_011514872.1 | 24 - 25 | 2408 | 123 | ||
| XM_011516571.2 | XP_011514873.1 | 24 - 25 | 2388 | 123 | ||
| XM_017012588.1 | XP_016868077.1 | 23 - 24 | 3569 | 123 | ||
| GenBank mRNA | BC117468.1 | - | 23 - 24 | 1920 | 40126019 | 123 |
| BC117470.1 | - | 23 - 24 | 1920 | 40126021 | 123 | |
| M76559.1 | - | 23 - 24 | 1923 | 123 | ||
More Information
Updated Mapping Information
Set Membership: |
|

CACNA2D1
Genomic Map