Assay Details
| Gene Symbol: COMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011529886.1 | XP_011528188.1 | 1 - 2 | 60 | 60 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011529886.1 transcripts.
Set Membership: |
|

COMT
Genomic Map