| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008347.hg.1 |
ENST00000583051.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000263202.14 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000263202.14 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000263202.14 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000263202.14 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000263202.14 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000263202.14 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000263202.14 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000263202.14 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000263202.14 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000263202.14 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000263202.14 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000263202.14 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000263202.14 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000263202.14 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000263202.14 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000263202.14 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000263202.14 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000263202.14 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000399523.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000399523.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000399523.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000399523.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000399523.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000399523.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000399523.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000399523.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000399523.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000399523.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000399523.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000399523.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000399523.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000399523.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000399523.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000421968.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000421968.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000421968.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000421968.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000421968.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000421968.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000421968.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000421968.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000447868.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000447868.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000447868.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000447868.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000447868.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000447868.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000447868.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000447868.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000459854.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000459854.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000459854.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000459854.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000459854.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000459854.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000459854.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000459854.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000459854.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000459854.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000459854.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000459854.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000459854.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000459854.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000459854.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000459854.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000459854.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000459854.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000466373.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000466373.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000466373.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000466373.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000466373.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000466373.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000466373.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000466373.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000466373.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000466373.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000466373.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000466373.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000474226.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000474226.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000474226.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000484101.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000484101.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000484101.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000484101.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000484101.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000484101.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000484101.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000484101.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000489406.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000489406.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000489406.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000489406.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000489406.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000489406.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000489406.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000489406.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000494054.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000494054.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000494054.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000494054.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000494054.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000494054.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
ENST00000494054.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
NM_001035247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
NM_001035247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
NM_001035247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
NM_001035247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
NM_001035247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
NM_001035247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
NM_001035247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
NM_001035247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
NM_001035247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
NM_001035247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
NM_001035247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
NM_001035247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
NM_001035247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
NM_001035247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
NM_001035247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
NM_001035247.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
NM_005659.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
NM_005659.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
NM_005659.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
NM_005659.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
NM_005659.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
NM_005659.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
NM_005659.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
NM_005659.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
NM_005659.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
NM_005659.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
NM_005659.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
NM_005659.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
NM_005659.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
NM_005659.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
NM_005659.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
NM_005659.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007982.hg.1 |
NM_005659.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008347.hg.1 |
ENST00000583051.1 |
Ensembl |