Assay Details
| Gene Symbol: ARHGEF33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145451.2 | NP_001138923.2 | 5 - 6 | 590 | 104 | |
| GenBank mRNA | DV080125.1 | - | 3 - 4 | 305 | 104 | |
| DV080126.1 | - | 1 - 2 | 104 | |||
| DY654603.1 | - | 1 - 2 | 68 | 104 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02004960.hg.1
Set Membership: |
|
Panther Classification:
Molecular Function -
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02003694.hg.1 | TC02004960.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003694.hg.1 | ENST00000398800.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003694.hg.1 | ENST00000536934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003694.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000411874.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000411874.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000411874.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000411874.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000411874.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000411874.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000430382.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000430382.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000430382.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000433605.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000433605.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000433605.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000433605.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000433605.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000483305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000483305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000483305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000486958.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000486958.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000486958.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000486958.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000486958.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000486958.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000488692.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000488692.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000488692.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000488692.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000488692.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221538.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221538.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221538.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221538.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221538.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221538.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221538.hg.1 | TC0200016431.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000411874.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000411874.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000411874.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000411874.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000411874.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000411874.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000430382.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000430382.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000430382.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000433605.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000433605.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000433605.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000433605.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000433605.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000483305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000483305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000483305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000486958.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000486958.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000486958.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000486958.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000486958.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000486958.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000488692.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000488692.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000488692.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000488692.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000488692.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | NM_001145451.2 | Refseq |

ARHGEF33
Genomic Map