Assay Details
| Gene Symbol: ARHGEF33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145451.2 | NP_001138923.2 | 6 - 7 | 646 | 94 | |
| GenBank mRNA | DV080125.1 | - | 4 - 5 | 361 | 94 | |
| DY654603.1 | - | 2 - 3 | 124 | 94 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02004960.hg.1
Set Membership: |
|
Panther Classification:
Molecular Function -
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000411874.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000411874.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000411874.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000411874.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000411874.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000411874.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000430382.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000430382.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000430382.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000433605.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000433605.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000433605.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000433605.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000433605.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000483305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000483305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000483305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000486958.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000486958.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000486958.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000486958.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000486958.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000486958.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000488692.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000488692.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000488692.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000488692.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | ENST00000488692.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003695.hg.1 | TC02004960.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003695.hg.1 | ENST00000398800.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003695.hg.1 | ENST00000536934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003695.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000411874.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000411874.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000411874.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000411874.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000411874.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000411874.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000430382.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000430382.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000430382.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000433605.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000433605.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000433605.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000433605.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000433605.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000483305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000483305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000483305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000486958.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000486958.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000486958.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000486958.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000486958.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000486958.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000488692.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000488692.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000488692.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000488692.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | ENST00000488692.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016431.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221539.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221539.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221539.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221539.hg.1 | ENST00000398800.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221539.hg.1 | ENST00000409978.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221539.hg.1 | NM_001145451.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221539.hg.1 | TC0200016431.hg.1 | Clariom D TC |

ARHGEF33
Genomic Map