Assay Details
| Gene Symbol: CORO2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003389.3 | NP_003380.3 | 1 - 2 | 269 | 70 | |
| GenBank mRNA | BM822208.1 | - | 1 - 2 | 65 | 70 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001404.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_003389.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017487.hg.1 | TC09001404.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017487.hg.1 | ENST00000343933.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017487.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017487.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017487.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017489.hg.1 | TC09001404.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017489.hg.1 | ENST00000343933.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017489.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174748.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174748.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174748.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174748.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174748.hg.1 | TC0900010969.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900174746.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174746.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174746.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174746.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174746.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174746.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174746.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174746.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174746.hg.1 | TC0900010969.hg.1 | Clariom D TC |

CORO2A
Genomic Map