Assay Details
| Gene Symbol: CKAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286687.1 | NP_001273616.1 | 2 - 3 | 351 | 130 | |
| NM_018204.4 | NP_060674.3 | 2 - 3 | 351 | 130 | ||
| GenBank mRNA | AK022982.1 | - | 2 - 3 | 250 | 130 | |
| AY062261.1 | - | 2 - 3 | 272 | 130 | ||
| AY062262.1 | - | 2 - 3 | 272 | 130 | ||
| BC105806.1 | - | 2 - 3 | 230 | 130 | ||
| BC130296.1 | - | 2 - 3 | 199 | 40146327 | 130 | |
| BC136332.1 | - | 2 - 3 | 199 | 9020319 | 130 | |
| BM541375.1 | - | 2 - 3 | 356 | 130 | ||
More Information
Set Membership: |
|

CKAP2
Genomic Map