Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000324266.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000324266.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000324266.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000324266.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000324266.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000324266.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000324266.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000324266.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000324266.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000324266.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000324266.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000324266.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000324266.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000324266.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000324266.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000324266.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000324266.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000324266.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000382110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000382110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000382110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000382110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000382110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000382110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000382110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000382110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000382110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000382110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000382110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000382110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000382110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000382110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000382110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000382110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000382110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000411973.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000411973.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000411973.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000411973.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000415624.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000415624.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000415624.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000415624.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000415624.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000415624.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000415624.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000416918.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000416918.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000416918.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000416918.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000417243.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000417243.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000417243.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000417243.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000417243.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000417243.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000417243.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000417243.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000417243.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000417243.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000433382.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000433382.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000433382.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000433382.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000433382.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000433382.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000437733.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000437733.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000437733.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000437733.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000437733.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000441099.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000441099.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000441099.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000441099.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000441099.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000441983.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000441983.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000441983.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000441983.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000441983.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000441983.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000441983.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000452495.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000452495.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000452495.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000452495.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000457845.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000457845.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000457845.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000457845.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000457845.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000457845.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000461577.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000461577.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000461577.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000461577.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000461577.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000462983.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000462983.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000462983.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000469147.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000469147.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000469147.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000469147.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000469147.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000469400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000469400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000469400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000469400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000469400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000469400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000473348.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000473348.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000473348.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000473348.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000473348.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000479897.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000479897.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000479897.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000479897.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000479955.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000479955.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000482645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000482645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000482645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000482645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000482645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000482645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000489032.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000489032.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000489032.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000493792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000493792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000493792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000493792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000493792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000497597.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000497597.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000497597.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000497597.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000497597.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000497597.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000497597.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000497597.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
ENST00000497597.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
NM_016030.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
NM_016030.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
NM_016030.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
NM_016030.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
NM_016030.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
NM_016030.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
NM_016030.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
NM_016030.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
NM_016030.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
NM_016030.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
NM_016030.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
NM_016030.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
NM_016030.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
NM_016030.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
NM_016030.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
NM_016030.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
NM_016030.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006524.hg.1 |
NM_016030.5 |
Refseq |