Assay Details
| Gene Symbol: SLC12A7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006598.2 | NP_006589.2 | 12 - 13 | 1695 | 99 | |
| XM_005248231.3 | XP_005248288.1 | 12 - 13 | 1700 | 99 | ||
| XM_011513939.2 | XP_011512241.1 | 12 - 13 | 1789 | 99 | ||
| XM_011513940.2 | XP_011512242.1 | 12 - 13 | 1624 | 99 | ||
| XM_011513941.2 | XP_011512243.2 | 12 - 13 | 1719 | 99 | ||
| XM_017008958.1 | XP_016864447.1 | 12 - 13 | 1999 | 99 | ||
| XM_017008959.1 | XP_016864448.1 | 12 - 13 | 1651 | 99 | ||
| GenBank mRNA | AF105365.1 | - | 12 - 13 | 1633 | 99 | |
| AK024493.1 | - | 5 - 6 | 720 | 99 | ||
| AK024497.1 | - | 5 - 6 | 545 | 99 | ||
| BC098390.1 | - | 12 - 13 | 1673 | 5265743 | 99 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05001102.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05016014.hg.1 | TC05001102.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05016014.hg.1 | ENST00000264930.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05016014.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000504576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000504576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000504576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000504576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000504576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000510943.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000510943.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000510943.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000510943.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000513223.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000513223.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000513223.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000513223.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000513223.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000513223.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000513223.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000513223.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000513223.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000514994.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000514994.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000514994.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000514994.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000514994.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05016015.hg.1 | TC05001102.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05016015.hg.1 | ENST00000264930.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05016015.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500169430.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169430.hg.1 | ENST00000504576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169430.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500169430.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169430.hg.1 | ENST00000504576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169430.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500169430.hg.1 | TC0500009837.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000504576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000504576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000504576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000504576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000504576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000510943.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000510943.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000510943.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000510943.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000513223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000513223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000513223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000513223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000513223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000513223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000513223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000513223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000513223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000514994.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000514994.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000514994.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000514994.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000514994.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500169431.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169431.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500169431.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169431.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500169431.hg.1 | TC0500009837.hg.1 | Clariom D TC |

SLC12A7
Genomic Map