Assay Details
| Gene Symbol: PYCRL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023078.3 | NP_075566.2 | 3 - 4 | 404 | 92 | |
| GenBank mRNA | AK023914.1 | - | 3 - 4 | 364 | 92 | |
| AK222685.1 | - | 374 | 92 | |||
| AK225404.1 | - | 3 - 4 | 353 | 92 | ||
| AK294615.1 | - | 4 - 5 | 472 | 92 | ||
| AK295444.1 | - | 3 - 4 | 345 | 92 | ||
| AL833857.1 | - | 2 - 3 | 248 | 92 | ||
| BC007993.1 | - | 3 - 4 | 384 | 3533609 | 92 | |
| DA403767.1 | - | 3 - 4 | 404 | 92 | ||
| DA629534.1 | - | 3 - 4 | 402 | 92 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08001722.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK294615.1,AK295444.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000220966.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000220966.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000220966.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000220966.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000220966.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000220966.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000220966.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000220966.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000220966.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000220966.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000220966.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000220966.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000220966.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000220966.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000377579.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000377579.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000377579.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000377579.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000377579.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000377579.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000377579.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000377579.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000377579.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000433751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000433751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000433751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000433751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000433751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000433751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000433751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000433751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000433751.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000447926.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000447926.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000447926.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000447926.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000447926.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000447926.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000447926.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000447926.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000447926.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000447926.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000447926.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000447926.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000447926.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000462036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000462036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000462036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000462036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000462036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000462036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000482616.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000482616.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000482616.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000482616.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000482616.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000482616.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000495276.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000495276.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000495276.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000495276.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000495276.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000495276.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000495276.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000495276.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000495276.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000495276.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000495276.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000495276.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | ENST00000495276.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | NM_023078.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | NM_023078.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | NM_023078.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | NM_023078.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | NM_023078.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | NM_023078.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | NM_023078.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | NM_023078.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | NM_023078.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | NM_023078.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | NM_023078.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | NM_023078.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | NM_023078.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012146.hg.1 | NM_023078.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08022755.hg.1 | TC08001722.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08022755.hg.1 | ENST00000220966.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08022755.hg.1 | ENST00000377579.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08022755.hg.1 | ENST00000495276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08022755.hg.1 | NM_023078.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08022753.hg.1 | TC08001722.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08022753.hg.1 | ENST00000220966.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08022753.hg.1 | ENST00000377579.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08022753.hg.1 | ENST00000495276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08022753.hg.1 | NM_023078.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000220966.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000220966.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000220966.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000220966.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000220966.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000220966.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000220966.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000220966.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000220966.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000220966.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000220966.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000220966.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000220966.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000220966.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000377579.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000377579.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000377579.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000377579.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000377579.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000377579.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000377579.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000377579.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000377579.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000433751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000433751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000433751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000433751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000433751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000433751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000433751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000433751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000433751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000447926.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000447926.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000447926.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000447926.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000447926.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000447926.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000447926.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000447926.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000447926.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000447926.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000447926.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000447926.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000447926.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000462036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000462036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000462036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000462036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000462036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000462036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000482616.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000482616.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000482616.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000482616.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000482616.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000482616.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000495276.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000495276.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000495276.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000495276.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000495276.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000495276.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000495276.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000495276.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000495276.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000495276.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000495276.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000495276.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | ENST00000495276.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | NM_023078.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | NM_023078.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | NM_023078.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | NM_023078.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | NM_023078.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | NM_023078.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | NM_023078.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | NM_023078.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | NM_023078.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | NM_023078.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | NM_023078.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | NM_023078.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | NM_023078.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012146.hg.1 | NM_023078.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800182869.hg.1 | ENST00000220966.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800182869.hg.1 | ENST00000377579.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800182869.hg.1 | ENST00000433751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800182869.hg.1 | ENST00000447926.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800182869.hg.1 | ENST00000462036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800182869.hg.1 | ENST00000482616.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800182869.hg.1 | ENST00000495276.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800182869.hg.1 | NM_023078.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800182869.hg.1 | ENST00000220966.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800182869.hg.1 | ENST00000377579.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800182869.hg.1 | ENST00000433751.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800182869.hg.1 | ENST00000447926.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800182869.hg.1 | ENST00000462036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800182869.hg.1 | ENST00000482616.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800182869.hg.1 | ENST00000495276.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800182869.hg.1 | NM_023078.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800182869.hg.1 | TC0800012146.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800182870.hg.1 | ENST00000220966.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800182870.hg.1 | ENST00000377579.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800182870.hg.1 | ENST00000447926.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800182870.hg.1 | ENST00000462036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800182870.hg.1 | ENST00000482616.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800182870.hg.1 | ENST00000495276.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800182870.hg.1 | NM_023078.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800182870.hg.1 | ENST00000220966.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800182870.hg.1 | ENST00000377579.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800182870.hg.1 | ENST00000447926.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800182870.hg.1 | ENST00000462036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800182870.hg.1 | ENST00000482616.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800182870.hg.1 | ENST00000495276.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800182870.hg.1 | NM_023078.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800182870.hg.1 | TC0800012146.hg.1 | Clariom D TC |

PYCRL
Genomic Map