| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22000641.hg.1 |
ENST00000351989.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22000641.hg.1 |
ENST00000383024.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22000641.hg.1 |
ENST00000407755.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22000641.hg.1 |
NM_001190326.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22000641.hg.1 |
NM_022720.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22000641.hg.1 |
TC22000053.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR2200145529.hg.1 |
TC2200006636.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200145529.hg.1 |
ENST00000351989.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200145529.hg.1 |
ENST00000383024.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200145529.hg.1 |
ENST00000407755.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200145529.hg.1 |
ENST00000491892.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200145529.hg.1 |
ENST00000495826.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200145529.hg.1 |
ENST00000498171.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200145529.hg.1 |
NM_001190326.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200145529.hg.1 |
NM_022720.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200145529.hg.1 |
ENST00000351989.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200145529.hg.1 |
ENST00000383024.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200145529.hg.1 |
ENST00000407755.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200145529.hg.1 |
ENST00000491892.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200145529.hg.1 |
ENST00000495826.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200145529.hg.1 |
ENST00000498171.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200145529.hg.1 |
NM_001190326.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200145529.hg.1 |
NM_022720.6 |
Refseq |