Assay Details
| Gene Symbol: CNTNAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003632.2 | NP_003623.1 | 4 - 5 | 725 | 79 | |
| XM_005257748.4 | XP_005257805.1 | 4 - 5 | 1490 | 79 | ||
| XM_017025238.1 | XP_016880727.1 | 6 - 7 | 1860 | 79 | ||
| GenBank mRNA | U87223.1 | - | 4 - 5 | 726 | 79 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000533.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
protein localization to paranode region of axon
cytoskeleton organization
cell adhesion
signal transduction
positive regulation of signal transduction
neuronal action potential propagation
paranodal junction assembly
neuron projection morphogenesis
neuromuscular process controlling posture
neuromuscular process controlling balance
receptor activity
SH3/SH2 adaptor activity
protein binding
SH3 domain binding
protein localization to paranode region of axon
cytoskeleton organization
cell adhesion
signal transduction
positive regulation of signal transduction
neuronal action potential propagation
paranodal junction assembly
neuron projection morphogenesis
neuromuscular process controlling posture
neuromuscular process controlling balance
receptor activity
SH3/SH2 adaptor activity
protein binding
SH3 domain binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17006470.hg.1 | ENST00000264638.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006470.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006470.hg.1 | TC17000533.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000585534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000585534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000585534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000586801.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000586801.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000586801.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000591662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000591662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000591662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000591662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000591662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000591662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000591662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000591662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000591662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000591662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000591662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000591662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000591662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000591662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000591662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000591662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000591662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000591662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000591662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000591662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000591662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000591662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000591662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000591662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000591662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000591662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | ENST00000591662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700157641.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157641.hg.1 | ENST00000591662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157641.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700157641.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157641.hg.1 | ENST00000591662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157641.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700157641.hg.1 | TC1700007910.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000264638.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000585534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000585534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000585534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000586801.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000586801.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000586801.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000591662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000591662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000591662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000591662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000591662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000591662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000591662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000591662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000591662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000591662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000591662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000591662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000591662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000591662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000591662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000591662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000591662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000591662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000591662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000591662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000591662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000591662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000591662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000591662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000591662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000591662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | ENST00000591662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | NM_003632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007910.hg.1 | NM_003632.2 | Refseq |

CNTNAP1
Genomic Map