Assay Details
Gene Symbol: SPATC1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142854.1 | NP_001136326.1 | 2 - 3 | 1257 | 158 | |
XM_005261188.4 | XP_005261245.1 | 2 - 3 | 1032 | 158 | ||
XM_017028480.1 | XP_016883969.1 | 3 - 4 | 979 | 158 | ||
GenBank mRNA | AK304535.1 | - | 2 - 3 | 1257 | 158 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC21000553.hg.1
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC2100008461.hg.1 | ENST00000291672.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008461.hg.1 | ENST00000291672.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008461.hg.1 | ENST00000291672.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008461.hg.1 | ENST00000291672.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008461.hg.1 | ENST00000291672.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008461.hg.1 | ENST00000291672.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008461.hg.1 | ENST00000291672.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008461.hg.1 | ENST00000291672.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008461.hg.1 | ENST00000291672.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008461.hg.1 | ENST00000291672.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008461.hg.1 | ENST00000291672.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008461.hg.1 | ENST00000291672.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008461.hg.1 | ENST00000330205.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008461.hg.1 | ENST00000330205.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008461.hg.1 | ENST00000330205.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008461.hg.1 | ENST00000330205.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008461.hg.1 | ENST00000330205.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008461.hg.1 | ENST00000330205.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008461.hg.1 | ENST00000330205.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008461.hg.1 | ENST00000330205.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008461.hg.1 | ENST00000330205.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008461.hg.1 | ENST00000330205.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008461.hg.1 | NM_001142854.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008461.hg.1 | NM_001142854.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008461.hg.1 | NM_001142854.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008461.hg.1 | NM_001142854.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008461.hg.1 | NM_001142854.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008461.hg.1 | NM_001142854.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008461.hg.1 | NM_001142854.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008461.hg.1 | NM_001142854.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008461.hg.1 | NM_001142854.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008461.hg.1 | NM_001142854.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008461.hg.1 | NM_001142854.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008461.hg.1 | NM_001142854.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008461.hg.1 | NM_032261.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008461.hg.1 | NM_032261.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008461.hg.1 | NM_032261.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008461.hg.1 | NM_032261.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008461.hg.1 | NM_032261.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008461.hg.1 | NM_032261.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008461.hg.1 | NM_032261.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008461.hg.1 | NM_032261.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008461.hg.1 | NM_032261.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008461.hg.1 | NM_032261.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR21006673.hg.1 | ENST00000291672.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR21006673.hg.1 | ENST00000330205.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR21006673.hg.1 | NM_001142854.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR21006673.hg.1 | NM_032261.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR21006673.hg.1 | TC21000553.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR21006676.hg.1 | ENST00000291672.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR21006676.hg.1 | NM_001142854.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR21006676.hg.1 | TC21000553.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR21006674.hg.1 | ENST00000291672.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR21006674.hg.1 | ENST00000330205.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR21006674.hg.1 | NM_001142854.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR21006674.hg.1 | NM_032261.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR21006674.hg.1 | TC21000553.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR21006672.hg.1 | ENST00000291672.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR21006672.hg.1 | ENST00000330205.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR21006672.hg.1 | NM_001142854.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR21006672.hg.1 | NM_032261.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR21006672.hg.1 | TC21000553.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR2100157341.hg.1 | TC2100008461.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2100157341.hg.1 | ENST00000291672.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100157341.hg.1 | NM_001142854.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100157341.hg.1 | ENST00000291672.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100157341.hg.1 | NM_001142854.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100157340.hg.1 | TC2100008461.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2100157340.hg.1 | ENST00000291672.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100157340.hg.1 | NM_001142854.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100157340.hg.1 | ENST00000291672.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100157340.hg.1 | NM_001142854.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100157338.hg.1 | TC2100008461.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2100157338.hg.1 | ENST00000291672.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100157338.hg.1 | ENST00000330205.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100157338.hg.1 | NM_001142854.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100157338.hg.1 | NM_032261.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100157338.hg.1 | ENST00000291672.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100157338.hg.1 | ENST00000330205.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100157338.hg.1 | NM_001142854.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100157338.hg.1 | NM_032261.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100157336.hg.1 | TC2100008461.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2100157336.hg.1 | ENST00000291672.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100157336.hg.1 | ENST00000330205.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100157336.hg.1 | NM_001142854.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100157336.hg.1 | NM_032261.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100157336.hg.1 | ENST00000291672.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100157336.hg.1 | ENST00000330205.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100157336.hg.1 | NM_001142854.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100157336.hg.1 | NM_032261.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100157337.hg.1 | TC2100008461.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2100157337.hg.1 | ENST00000291672.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100157337.hg.1 | ENST00000330205.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100157337.hg.1 | NM_001142854.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100157337.hg.1 | NM_032261.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100157337.hg.1 | ENST00000291672.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100157337.hg.1 | ENST00000330205.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100157337.hg.1 | NM_001142854.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100157337.hg.1 | NM_032261.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008461.hg.1 | ENST00000291672.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008461.hg.1 | ENST00000291672.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008461.hg.1 | ENST00000291672.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008461.hg.1 | ENST00000291672.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008461.hg.1 | ENST00000291672.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008461.hg.1 | ENST00000291672.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008461.hg.1 | ENST00000291672.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008461.hg.1 | ENST00000291672.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008461.hg.1 | ENST00000291672.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008461.hg.1 | ENST00000291672.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008461.hg.1 | ENST00000291672.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008461.hg.1 | ENST00000291672.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008461.hg.1 | ENST00000330205.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008461.hg.1 | ENST00000330205.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008461.hg.1 | ENST00000330205.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008461.hg.1 | ENST00000330205.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008461.hg.1 | ENST00000330205.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008461.hg.1 | ENST00000330205.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008461.hg.1 | ENST00000330205.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008461.hg.1 | ENST00000330205.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008461.hg.1 | ENST00000330205.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008461.hg.1 | ENST00000330205.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008461.hg.1 | NM_001142854.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008461.hg.1 | NM_001142854.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008461.hg.1 | NM_001142854.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008461.hg.1 | NM_001142854.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008461.hg.1 | NM_001142854.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008461.hg.1 | NM_001142854.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008461.hg.1 | NM_001142854.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008461.hg.1 | NM_001142854.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008461.hg.1 | NM_001142854.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008461.hg.1 | NM_001142854.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008461.hg.1 | NM_001142854.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008461.hg.1 | NM_001142854.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008461.hg.1 | NM_032261.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008461.hg.1 | NM_032261.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008461.hg.1 | NM_032261.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008461.hg.1 | NM_032261.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008461.hg.1 | NM_032261.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008461.hg.1 | NM_032261.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008461.hg.1 | NM_032261.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008461.hg.1 | NM_032261.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008461.hg.1 | NM_032261.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008461.hg.1 | NM_032261.4 | Refseq |
