Assay Details
| Gene Symbol: TPPP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016140.3 | NP_057224.2 | 2 - 3 | 246 | 82 | |
| GenBank mRNA | BI754230.1 | - | 2 - 3 | 229 | 82 | |
| BI772576.1 | - | 1 - 2 | 524 | 82 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16001187.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_016140.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000290942.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000290942.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000290942.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000290942.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000290942.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000290942.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000290942.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000393957.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000393957.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000393957.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000393957.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000393957.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000393957.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000458950.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000561537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000561537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000561537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000561537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000561537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000561537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000562206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000562206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000562206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000562206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000562206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000562206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000562206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000564104.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000564104.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000564104.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000564104.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000564104.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | NM_015964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | NM_015964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | NM_015964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | NM_015964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | NM_015964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | NM_015964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | NM_016140.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | NM_016140.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | NM_016140.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | NM_016140.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | NM_016140.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | NM_016140.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | NM_016140.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016522.hg.1 | ENST00000290942.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016522.hg.1 | ENST00000393957.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016522.hg.1 | NM_015964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016522.hg.1 | NM_016140.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016522.hg.1 | TC16001187.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016524.hg.1 | ENST00000290942.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016524.hg.1 | NM_016140.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016524.hg.1 | TC16001187.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1600181900.hg.1 | ENST00000290942.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181900.hg.1 | ENST00000562206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181900.hg.1 | NM_016140.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600181900.hg.1 | ENST00000290942.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181900.hg.1 | ENST00000562206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181900.hg.1 | NM_016140.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600181900.hg.1 | TC1600010627.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600181897.hg.1 | ENST00000290942.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181897.hg.1 | ENST00000393957.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181897.hg.1 | ENST00000561537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181897.hg.1 | ENST00000562206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181897.hg.1 | ENST00000564104.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181897.hg.1 | NM_015964.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600181897.hg.1 | NM_016140.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600181897.hg.1 | ENST00000290942.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181897.hg.1 | ENST00000393957.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181897.hg.1 | ENST00000561537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181897.hg.1 | ENST00000562206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181897.hg.1 | ENST00000564104.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181897.hg.1 | NM_015964.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600181897.hg.1 | NM_016140.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600181897.hg.1 | TC1600010627.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000290942.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000290942.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000290942.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000290942.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000290942.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000290942.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000290942.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000393957.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000393957.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000393957.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000393957.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000393957.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000393957.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000458950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000561537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000561537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000561537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000561537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000561537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000561537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000562206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000562206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000562206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000562206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000562206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000562206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000562206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000564104.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000564104.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000564104.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000564104.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000564104.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | NM_015964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | NM_015964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | NM_015964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | NM_015964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | NM_015964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | NM_015964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | NM_016140.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | NM_016140.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | NM_016140.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | NM_016140.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | NM_016140.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | NM_016140.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | NM_016140.3 | Refseq |

TPPP3
Genomic Map