Assay Details
| Gene Symbol: DIO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007023.4 | NP_001007024.1 | 2 - 3 | 482 | 136 | |
| GenBank mRNA | AB041843.1 | - | 2 - 3 | 355 | 136 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14001364.hg.1
Set Membership: |
|

DIO2
Genomic Map