Assay Details
| Gene Symbol: IL10RB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000628.4 | NP_000619.3 | 1 - 2 | 155 | 97 | |
| GenBank mRNA | AK313348.1 | - | 1 - 2 | 148 | 97 | |
| AU133530.2 | - | 1 - 2 | 155 | 97 | ||
| BC001903.1 | - | 1 - 2 | 134 | 3544611 | 97 | |
| Z17227.1 | - | 1 - 2 | 92 | 97 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC21001059.hg
Additional Information:
This assay is in the 5'UTR region of the BC001903.1,NM_000628.4,Z17227.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR21000875.hg.1 | TC21001059.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000875.hg.1 | ENST00000290200.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000875.hg.1 | NM_000628.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000876.hg.1 | TC21001059.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000876.hg.1 | ENST00000290200.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000876.hg.1 | ENST00000539894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000876.hg.1 | NM_000628.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000879.hg.1 | TC21001059.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000879.hg.1 | ENST00000290200.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000879.hg.1 | ENST00000539894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000879.hg.1 | NM_000628.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008495.hg.1 | ENST00000290200.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008495.hg.1 | ENST00000290200.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008495.hg.1 | ENST00000290200.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008495.hg.1 | ENST00000290200.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008495.hg.1 | ENST00000290200.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008495.hg.1 | ENST00000290200.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008495.hg.1 | ENST00000290200.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008495.hg.1 | ENST00000290200.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008495.hg.1 | ENST00000290200.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008495.hg.1 | ENST00000290200.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008495.hg.1 | ENST00000290200.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008495.hg.1 | ENST00000422891.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008495.hg.1 | ENST00000422891.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008495.hg.1 | ENST00000422891.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008495.hg.1 | ENST00000422891.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008495.hg.1 | ENST00000422891.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008495.hg.1 | ENST00000422891.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008495.hg.1 | ENST00000422891.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008495.hg.1 | ENST00000451065.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008495.hg.1 | ENST00000451065.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008495.hg.1 | ENST00000451065.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008495.hg.1 | ENST00000451065.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008495.hg.1 | ENST00000451065.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008495.hg.1 | ENST00000451065.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008495.hg.1 | ENST00000451065.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008495.hg.1 | ENST00000493295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008495.hg.1 | ENST00000493295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008495.hg.1 | ENST00000493295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008495.hg.1 | ENST00000493295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008495.hg.1 | ENST00000493295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008495.hg.1 | ENST00000493295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008495.hg.1 | ENST00000493295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008495.hg.1 | ENST00000493295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008495.hg.1 | ENST00000493295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008495.hg.1 | ENST00000493295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008495.hg.1 | ENST00000498371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008495.hg.1 | ENST00000498371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008495.hg.1 | ENST00000498371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008495.hg.1 | ENST00000498371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008495.hg.1 | NM_000628.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008495.hg.1 | NM_000628.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008495.hg.1 | NM_000628.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008495.hg.1 | NM_000628.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008495.hg.1 | NM_000628.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008495.hg.1 | NM_000628.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008495.hg.1 | NM_000628.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008495.hg.1 | NM_000628.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008495.hg.1 | NM_000628.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008495.hg.1 | NM_000628.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008495.hg.1 | NM_000628.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100157913.hg.1 | TC2100008495.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100157913.hg.1 | ENST00000290200.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100157913.hg.1 | ENST00000422891.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100157913.hg.1 | NM_000628.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100157913.hg.1 | ENST00000290200.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100157913.hg.1 | ENST00000422891.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100157913.hg.1 | NM_000628.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008495.hg.1 | ENST00000290200.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008495.hg.1 | ENST00000290200.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008495.hg.1 | ENST00000290200.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008495.hg.1 | ENST00000290200.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008495.hg.1 | ENST00000290200.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008495.hg.1 | ENST00000290200.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008495.hg.1 | ENST00000290200.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008495.hg.1 | ENST00000290200.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008495.hg.1 | ENST00000290200.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008495.hg.1 | ENST00000290200.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008495.hg.1 | ENST00000290200.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008495.hg.1 | ENST00000422891.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008495.hg.1 | ENST00000422891.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008495.hg.1 | ENST00000422891.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008495.hg.1 | ENST00000422891.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008495.hg.1 | ENST00000422891.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008495.hg.1 | ENST00000422891.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008495.hg.1 | ENST00000422891.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008495.hg.1 | ENST00000451065.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008495.hg.1 | ENST00000451065.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008495.hg.1 | ENST00000451065.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008495.hg.1 | ENST00000451065.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008495.hg.1 | ENST00000451065.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008495.hg.1 | ENST00000451065.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008495.hg.1 | ENST00000451065.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008495.hg.1 | ENST00000493295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008495.hg.1 | ENST00000493295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008495.hg.1 | ENST00000493295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008495.hg.1 | ENST00000493295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008495.hg.1 | ENST00000493295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008495.hg.1 | ENST00000493295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008495.hg.1 | ENST00000493295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008495.hg.1 | ENST00000493295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008495.hg.1 | ENST00000493295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008495.hg.1 | ENST00000493295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008495.hg.1 | ENST00000498371.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008495.hg.1 | ENST00000498371.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008495.hg.1 | ENST00000498371.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008495.hg.1 | ENST00000498371.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008495.hg.1 | NM_000628.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008495.hg.1 | NM_000628.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008495.hg.1 | NM_000628.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008495.hg.1 | NM_000628.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008495.hg.1 | NM_000628.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008495.hg.1 | NM_000628.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008495.hg.1 | NM_000628.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008495.hg.1 | NM_000628.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008495.hg.1 | NM_000628.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008495.hg.1 | NM_000628.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008495.hg.1 | NM_000628.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100157912.hg.1 | TC2100008495.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100157912.hg.1 | ENST00000290200.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100157912.hg.1 | ENST00000422891.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100157912.hg.1 | NM_000628.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100157912.hg.1 | ENST00000290200.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100157912.hg.1 | ENST00000422891.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100157912.hg.1 | NM_000628.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100157916.hg.1 | TC2100008495.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100157916.hg.1 | ENST00000290200.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100157916.hg.1 | ENST00000422891.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100157916.hg.1 | ENST00000493295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100157916.hg.1 | ENST00000498371.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100157916.hg.1 | NM_000628.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100157916.hg.1 | ENST00000290200.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100157916.hg.1 | ENST00000422891.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100157916.hg.1 | ENST00000493295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100157916.hg.1 | ENST00000498371.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100157916.hg.1 | NM_000628.4 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 200014 | - | - |

IL10RB
Genomic Map