Assay Details
| Gene Symbol: TTC7B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320421.1 | NP_001307350.1 | 18 - 19 | 2142 | 80 | |
| XM_005267367.1 | XP_005267424.1 | 15 - 16 | 1619 | 80 | ||
| XM_011536497.1 | XP_011534799.1 | 19 - 20 | 2366 | 80 | ||
| XM_017021044.1 | XP_016876533.1 | 19 - 20 | 2272 | 80 | ||
| XM_017021045.1 | XP_016876534.1 | 18 - 19 | 2110 | 80 | ||
| GenBank mRNA | BX247966.1 | - | 4 - 5 | 434 | 80 | |
More Information
Set Membership: |
|

TTC7B
Genomic Map