Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009339.hg.1 |
TC06000782.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009339.hg.1 |
ENST00000305344.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009339.hg.1 |
NM_000865.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009340.hg.1 |
TC06000782.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009340.hg.1 |
ENST00000305344.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009340.hg.1 |
ENST00000369584.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009340.hg.1 |
NM_000865.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009344.hg.1 |
TC06000782.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009344.hg.1 |
ENST00000305344.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009344.hg.1 |
ENST00000369584.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009344.hg.1 |
NM_000865.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009343.hg.1 |
TC06000782.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009343.hg.1 |
ENST00000305344.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009343.hg.1 |
ENST00000369584.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009343.hg.1 |
NM_000865.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008716.hg.1 |
ENST00000305344.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008716.hg.1 |
ENST00000305344.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008716.hg.1 |
ENST00000305344.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008716.hg.1 |
ENST00000305344.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008716.hg.1 |
ENST00000305344.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008716.hg.1 |
ENST00000305344.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008716.hg.1 |
NM_000865.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008716.hg.1 |
NM_000865.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008716.hg.1 |
NM_000865.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008716.hg.1 |
NM_000865.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008716.hg.1 |
NM_000865.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008716.hg.1 |
NM_000865.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009341.hg.1 |
TC06000782.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009341.hg.1 |
ENST00000305344.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009341.hg.1 |
ENST00000369584.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009341.hg.1 |
NM_000865.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600160483.hg.1 |
ENST00000305344.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600160483.hg.1 |
NM_000865.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600160483.hg.1 |
ENST00000305344.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600160483.hg.1 |
NM_000865.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600160483.hg.1 |
TC0600008716.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600160485.hg.1 |
ENST00000305344.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600160485.hg.1 |
NM_000865.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600160485.hg.1 |
ENST00000305344.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600160485.hg.1 |
NM_000865.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600160485.hg.1 |
TC0600008716.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600160484.hg.1 |
ENST00000305344.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600160484.hg.1 |
NM_000865.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600160484.hg.1 |
ENST00000305344.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600160484.hg.1 |
NM_000865.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600160484.hg.1 |
TC0600008716.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600160486.hg.1 |
ENST00000305344.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600160486.hg.1 |
NM_000865.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600160486.hg.1 |
ENST00000305344.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600160486.hg.1 |
NM_000865.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600160486.hg.1 |
TC0600008716.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600008716.hg.1 |
ENST00000305344.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008716.hg.1 |
ENST00000305344.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008716.hg.1 |
ENST00000305344.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008716.hg.1 |
ENST00000305344.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008716.hg.1 |
ENST00000305344.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008716.hg.1 |
ENST00000305344.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008716.hg.1 |
NM_000865.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008716.hg.1 |
NM_000865.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008716.hg.1 |
NM_000865.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008716.hg.1 |
NM_000865.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008716.hg.1 |
NM_000865.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008716.hg.1 |
NM_000865.2 |
Refseq |