Assay Details
| Gene Symbol: PLBD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159727.1 | NP_001153199.1 | 1 - 2 | 321 | 60 | |
| NM_173542.3 | NP_775813.2 | 1 - 2 | 321 | 60 | ||
| GenBank mRNA | BC030618.2 | - | 1 - 2 | 314 | 4822591 | 60 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000891.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000547163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000547163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000547163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000548997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000548997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000548997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000548997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011815.hg.1 | ENST00000280800.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011815.hg.1 | ENST00000545182.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011815.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011815.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011815.hg.1 | TC12000891.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011814.hg.1 | ENST00000280800.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011814.hg.1 | ENST00000545182.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011814.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011814.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011814.hg.1 | TC12000891.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1200166510.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166510.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166510.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200166510.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200166510.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166510.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166510.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200166510.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200166510.hg.1 | TC1200008942.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200166509.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166509.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166509.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200166509.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200166509.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166509.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166509.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200166509.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200166509.hg.1 | TC1200008942.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200166511.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166511.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166511.hg.1 | ENST00000548997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166511.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200166511.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200166511.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166511.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166511.hg.1 | ENST00000548997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166511.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200166511.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200166511.hg.1 | TC1200008942.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000547163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000547163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000547163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000548997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000548997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000548997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000548997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200166512.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166512.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166512.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200166512.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200166512.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166512.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166512.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200166512.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200166512.hg.1 | TC1200008942.hg.1 | Clariom D TC |

PLBD2
Genomic Map