Assay Details
| Gene Symbol: PLBD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159727.1 | NP_001153199.1 | 9 - 10 | 1373 | 73 | |
| NM_173542.3 | NP_775813.2 | 10 - 11 | 1469 | 73 | ||
| XM_011538023.2 | XP_011536325.1 | 9 - 10 | 1252 | 73 | ||
| XM_017018977.1 | XP_016874466.1 | 9 - 10 | 1365 | 73 | ||
| GenBank mRNA | BC030618.2 | - | 10 - 11 | 1462 | 4822591 | 73 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000891.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12011827.hg.1 | ENST00000280800.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011827.hg.1 | ENST00000545182.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011827.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011827.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011827.hg.1 | TC12000891.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000547163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000547163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000547163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000548997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000548997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000548997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | ENST00000548997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200166525.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166525.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166525.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200166525.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200166525.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166525.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166525.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200166525.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200166525.hg.1 | TC1200008942.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000280800.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000545182.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000547163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000547163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000547163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000548997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000548997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000548997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | ENST00000548997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_001159727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_173542.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008942.hg.1 | NM_173542.3 | Refseq |

PLBD2
Genomic Map