Assay Details
| Gene Symbol: GDF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020634.1 | NP_065685.1 | 1 - 2 | 304 | 76 | |
| GenBank mRNA | AF110641.1 | - | 1 - 2 | 282 | 76 | |
| AF263538.1 | - | 1 - 2 | 304 | 76 | ||
| AY358875.1 | - | 1 - 2 | 333 | 76 | ||
| BC030959.1 | - | 1 - 2 | 316 | 4610141 | 76 | |
More Information
Set Membership: |
|

GDF3
Genomic Map