Assay Details
| Gene Symbol: PRTFDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282786.1 | NP_001269715.1 | 4 - 5 | 470 | 66 | |
| NM_020200.6 | NP_064585.1 | 4 - 5 | 470 | 66 | ||
| GenBank mRNA | AB209792.1 | - | 4 - 5 | 420 | 66 | |
| AF226056.1 | - | 4 - 5 | 418 | 66 | ||
| AK021950.1 | - | 4 - 5 | 430 | 66 | ||
| AK222674.1 | - | 4 - 5 | 429 | 66 | ||
| AK294130.1 | - | 4 - 5 | 429 | 66 | ||
| BC008662.2 | - | 4 - 5 | 432 | 3871911 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

PRTFDC1
Genomic Map