Assay Details
| Gene Symbol: CLCA3P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_024604.1 | 10 - 11 | 1539 | 141 | ||
| GenBank mRNA | AF043976.1 | - | 10 - 11 | 1539 | 141 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CLCA3P| Gene Symbol: CLCA3P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_024604.1 | 10 - 11 | 1539 | 141 | ||
| GenBank mRNA | AF043976.1 | - | 10 - 11 | 1539 | 141 | |
Set Membership: |
|