Assay Details
| Gene Symbol: CLCA3P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_024604.1 | 14 - 15 | 2437 | 122 | ||
| GenBank mRNA | AF043976.1 | - | 14 - 15 | 2438 | 122 | |
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
proteolysis
transport
ion transmembrane transport
regulation of ion transmembrane transport
calcium ion transmembrane transport
chloride transmembrane transport
metalloendopeptidase activity
transporter activity
voltage-gated ion channel activity
chloride channel activity
calcium channel activity
proteolysis
transport
ion transmembrane transport
regulation of ion transmembrane transport
calcium ion transmembrane transport
chloride transmembrane transport
metalloendopeptidase activity
transporter activity
voltage-gated ion channel activity
chloride channel activity
calcium channel activity
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 51438 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 77945 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 8x60K | 12067 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 966906 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 777092 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 381412 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 744219 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 628392 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 266223 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 530398 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 312182 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P34382 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 41247 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 27505 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_14_P139608 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P35210945 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P56139138 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P34382 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_23_P34382 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20381510 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00352344 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00352339 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20381496 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20381506 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20381508 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20381493 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00352352 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20381505 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20381501 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20381497 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20381500 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20381503 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20381510 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20381497 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00352339 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00352344 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20381505 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00352352 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20381508 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20381500 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20381501 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20381493 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20381496 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20381506 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20381503 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P34382 | - | - |
| Agilent Human Genome CGH Microarray 4x44K | A_14_P139608 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P34382 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352454 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352384 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352451 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352463 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352398 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352356 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15081543 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15081535 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15081537 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352468 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352433 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352409 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352390 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352346 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352479 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352395 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352393 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352350 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352426 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352386 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352414 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352373 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352461 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352465 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352342 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352343 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352378 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352457 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352416 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352428 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352481 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352369 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352445 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352472 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352406 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352402 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15081540 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352401 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352365 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352382 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352359 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352449 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352368 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352375 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352441 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352452 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352470 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352352 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15081533 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352437 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352371 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15081538 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352436 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352477 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352345 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352418 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15081547 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352363 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15081539 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15081548 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352420 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352358 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352405 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352423 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352439 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352483 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352354 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352475 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00352412 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P35210922 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P35210945 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_14_P139608 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P15201948 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 7902765 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7902758 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7902755 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7902757 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7902764 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7902759 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7902763 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7902770 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7902761 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7902766 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7902762 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7902760 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7902767 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7902756 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7902768 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7902769 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7902754 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 220810_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11750600_PM_x_at | - | - |
| Affymetrix Human Genome U219 Array | 11750600_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 220810_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 220810_at | - | - |
| Human Genome U133A 2.0 Array | 220810_PM_at | - | - |
| Human Genome U133A 2.0 Array | 220810_at | - | - |
| AffymetrixPathwork Diagnostics Pathchip | pw584928 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220810_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220810_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220810_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220810_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220810_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220810_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220810_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220810_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220810_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220810_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 220810_at9 | - | - |
| Human Genome U133 Set | 220810_PM_at | - | - |
| Human Genome U133 Set | 220810_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A007358 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A025427 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A025427 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A007358 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1744969 | - | - |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1773681 | AF043976.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1773681 | NR_024604.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1744969 | - | - |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1744969 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1773681 | AF043976.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1773681 | NR_024604.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1773681 | AF043976.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1773681 | NR_024604.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1773681 | AF043976.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1773681 | NR_024604.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1773681 | AF043976.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1773681 | NR_024604.1 | human_transcriptome |

CLCA3P
Genomic Map