Assay Details
| Gene Symbol: CLCA3P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_024604.1 | 8 - 9 | 1378 | 132 | ||
| GenBank mRNA | AF043976.1 | - | 8 - 9 | 1378 | 132 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000825.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
proteolysis
transport
ion transmembrane transport
regulation of ion transmembrane transport
calcium ion transmembrane transport
chloride transmembrane transport
metalloendopeptidase activity
transporter activity
voltage-gated ion channel activity
chloride channel activity
calcium channel activity
proteolysis
transport
ion transmembrane transport
regulation of ion transmembrane transport
calcium ion transmembrane transport
chloride transmembrane transport
metalloendopeptidase activity
transporter activity
voltage-gated ion channel activity
chloride channel activity
calcium channel activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01013131.hg.1 | TC01000825.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013131.hg.1 | ENST00000466454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01013131.hg.1 | NR_024604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000284054.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000284054.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000284054.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000284054.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000284054.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000284054.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000284054.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000284054.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000284054.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000284054.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000284054.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000284054.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000284054.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | NR_024604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | NR_024604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | NR_024604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | NR_024604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | NR_024604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | NR_024604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | NR_024604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | NR_024604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | NR_024604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | NR_024604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | NR_024604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | NR_024604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | NR_024604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | NR_024604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | NR_024604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | NR_024604.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100163693.hg.1 | ENST00000466454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100163693.hg.1 | ENST00000490028.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100163693.hg.1 | NR_024604.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100163693.hg.1 | ENST00000466454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100163693.hg.1 | ENST00000490028.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100163693.hg.1 | NR_024604.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100163693.hg.1 | TC0100008925.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000284054.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000284054.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000284054.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000284054.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000284054.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000284054.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000284054.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000284054.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000284054.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000284054.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000284054.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000284054.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000284054.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | NR_024604.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | NR_024604.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | NR_024604.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | NR_024604.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | NR_024604.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | NR_024604.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | NR_024604.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | NR_024604.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | NR_024604.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | NR_024604.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | NR_024604.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | NR_024604.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | NR_024604.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | NR_024604.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | NR_024604.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | NR_024604.1 | Refseq |

CLCA3P
Genomic Map