Assay Details
Gene Symbol: GRM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000844.3 | NP_000835.1 | 4 - 5 | 1303 | 126 | |
NM_181874.2 | NP_870989.1 | 4 - 5 | 1303 | 126 | ||
XM_017006272.1 | XP_016861761.1 | 4 - 5 | 769 | 126 | ||
XM_017006273.1 | XP_016861762.1 | 4 - 5 | 769 | 126 | ||
GenBank mRNA | AB209214.1 | - | 2 - 3 | 294 | 126 | |
AK312488.1 | - | 4 - 5 | 1303 | 126 | ||
BC136458.1 | - | 4 - 5 | 1119 | 9020447 | 126 | |
BC136459.1 | - | 4 - 5 | 1119 | 9020448 | 126 | |
X94552.1 | - | 4 - 5 | 1267 | 126 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC03000024.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
conditioned taste aversion
behavioral fear response
adenylate cyclase-inhibiting G-protein coupled glutamate receptor signaling pathway
chemical synaptic transmission
sensory perception of sound
sensory perception of smell
short-term memory
negative regulation of glutamate secretion
transmission of nerve impulse
adult behavior
negative regulation of cAMP biosynthetic process
regulation of cyclase activity
regulation of synaptic transmission, glutamatergic
calcium ion transmembrane transport
group III metabotropic glutamate receptor activity
voltage-gated calcium channel activity
calcium channel regulator activity
glutamate receptor activity
adenylate cyclase inhibitor activity
serine binding
conditioned taste aversion
behavioral fear response
adenylate cyclase-inhibiting G-protein coupled glutamate receptor signaling pathway
chemical synaptic transmission
sensory perception of sound
sensory perception of smell
short-term memory
negative regulation of glutamate secretion
transmission of nerve impulse
adult behavior
negative regulation of cAMP biosynthetic process
regulation of cyclase activity
regulation of synaptic transmission, glutamatergic
calcium ion transmembrane transport
group III metabotropic glutamate receptor activity
voltage-gated calcium channel activity
calcium channel regulator activity
glutamate receptor activity
adenylate cyclase inhibitor activity
serine binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000357716.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000357716.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000357716.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000357716.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000357716.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000357716.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000357716.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000357716.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000357716.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000357716.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000357716.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000357716.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000357716.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000357716.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000357716.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000357716.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000389335.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000389335.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000389335.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000389335.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000389335.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000389335.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000389335.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000389335.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000389335.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000389335.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000389335.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000389335.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000389335.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000389335.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000389335.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000389336.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000389336.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000389336.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000389336.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000389336.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000389336.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000389336.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000389336.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000389336.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000389336.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000389336.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000389336.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000389336.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000389336.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000402647.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000402647.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000402647.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000402647.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000402647.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000402647.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000402647.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000402647.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000402647.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000402647.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000435689.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000435689.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000435689.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000435689.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000435689.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000435689.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000440923.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000440923.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000440923.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000440923.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000440923.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000440923.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000440923.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000440923.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000440923.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000440923.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000440923.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000440923.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000440923.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000440923.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000440923.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000440923.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000443259.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000443259.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000443259.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000443259.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000443259.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000443259.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000445087.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000445087.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000445087.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000445087.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000445087.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000448328.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000448328.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000448328.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000448328.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000448328.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000458641.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000458641.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000458641.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000458641.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000458641.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000458641.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000458641.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000458641.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000458641.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000461677.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000461677.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000461677.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000461677.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000461677.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000463676.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000463676.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000463676.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000463676.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000463676.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000463676.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000463676.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000463676.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000463676.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000463676.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000467425.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000467425.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000467425.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000467425.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000467425.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000467425.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000467425.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000467425.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000467425.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000467425.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000467425.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000467425.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000467425.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000467425.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000467425.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000467425.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000467425.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000467425.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000471242.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000471242.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000486284.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000486284.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000486284.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000486284.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000486284.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000486284.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000486284.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000486284.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000486284.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000486284.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000486284.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000486284.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000486284.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000486284.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000486284.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000486284.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | ENST00000486284.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | NM_000844.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | NM_000844.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | NM_000844.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | NM_000844.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | NM_000844.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | NM_000844.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | NM_000844.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | NM_000844.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | NM_000844.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | NM_000844.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | NM_000844.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | NM_000844.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | NM_000844.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | NM_000844.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | NM_000844.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | NM_000844.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | NM_181874.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | NM_181874.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | NM_181874.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | NM_181874.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | NM_181874.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | NM_181874.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | NM_181874.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | NM_181874.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | NM_181874.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | NM_181874.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | NM_181874.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | NM_181874.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | NM_181874.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | NM_181874.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | NM_181874.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | NM_181874.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300006513.hg.1 | NM_181874.2 | Refseq |
