Assay Details
| Gene Symbol: NOTUM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178493.5 | NP_848588.3 | 2 - 3 | 648 | 52 | |
| GenBank mRNA | AB062438.1 | - | 2 - 3 | 74 | 52 | |
| BC036872.1 | - | 2 - 3 | 197 | 5225889 | 52 | |
| BC060882.1 | - | 2 - 3 | 656 | 30336041 | 52 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17001968.hg
Additional Information:
This assay is in the 5'UTR region of the AB062438.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17026836.hg.1 | ENST00000409678.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17026836.hg.1 | NM_178493.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17026836.hg.1 | TC17001968.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000477214.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000477214.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000477214.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000477214.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000477214.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000477214.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000477214.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000477214.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000477214.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000489218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000489218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000489218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000489218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000489218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000489218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | NM_178493.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | NM_178493.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | NM_178493.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | NM_178493.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | NM_178493.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | NM_178493.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | NM_178493.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | NM_178493.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | NM_178493.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | NM_178493.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | NM_178493.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | NM_178493.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | NM_178493.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | NM_178493.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | NM_178493.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | NM_178493.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000477214.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000477214.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000477214.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000477214.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000477214.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000477214.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000477214.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000477214.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000477214.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000489218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000489218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000489218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000489218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000489218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000489218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | NM_178493.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | NM_178493.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | NM_178493.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | NM_178493.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | NM_178493.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | NM_178493.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | NM_178493.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | NM_178493.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | NM_178493.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | NM_178493.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | NM_178493.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | NM_178493.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | NM_178493.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | NM_178493.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | NM_178493.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | NM_178493.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700199075.hg.1 | ENST00000409678.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700199075.hg.1 | ENST00000425009.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700199075.hg.1 | ENST00000477214.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700199075.hg.1 | ENST00000489218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700199075.hg.1 | NM_178493.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700199075.hg.1 | ENST00000409678.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700199075.hg.1 | ENST00000425009.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700199075.hg.1 | ENST00000477214.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700199075.hg.1 | ENST00000489218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700199075.hg.1 | NM_178493.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700199075.hg.1 | TC1700012084.hg.1 | Clariom D TC |

NOTUM
Genomic Map