Assay Details
Gene Symbol: DHX34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014681.5 | NP_055496.2 | 13 - 14 | 3054 | 75 | |
XM_005259443.3 | XP_005259500.1 | 13 - 14 | 3565 | 75 | ||
XM_011527550.2 | XP_011525852.1 | 13 - 14 | 3689 | 75 | ||
XM_011527551.2 | XP_011525853.1 | 13 - 14 | 3003 | 75 | ||
XM_017027524.1 | XP_016883013.1 | 12 - 13 | 2968 | 75 | ||
GenBank mRNA | AK123046.1 | - | 5 - 6 | 1864 | 75 | |
AK297692.1 | - | 8 - 9 | 1213 | 75 | ||
AW886612.1 | - | 2 - 3 | 260 | 75 | ||
BF894697.1 | - | 4 - 5 | 486 | 75 | ||
BU858266.1 | - | 2 - 3 | 182 | 75 | ||
D50924.1 | - | 8 - 9 | 2105 | 75 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC19000678.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000471451.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000471451.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000471451.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000471451.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000471451.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19009492.hg.1 | ENST00000257252.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19009492.hg.1 | ENST00000328771.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19009492.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19009492.hg.1 | TC19000678.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1900164732.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900164732.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900164732.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900164732.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900164732.hg.1 | TC1900008420.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900164732.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900164732.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900164732.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900164732.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000471451.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000471451.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000471451.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000471451.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000471451.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
