Assay Details
| Gene Symbol: RTCA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130841.1 | NP_001124313.1 | 10 - 11 | 1158 | 104 | |
| NM_003729.3 | NP_003720.1 | 9 - 10 | 1119 | 104 | ||
| XM_005271298.4 | XP_005271355.1 | 9 - 10 | 1116 | 104 | ||
| XM_005271299.4 | XP_005271356.1 | 9 - 10 | 1105 | 104 | ||
| XM_017002669.1 | XP_016858158.1 | 9 - 10 | 862 | 104 | ||
| GenBank mRNA | AK307445.1 | - | 7 - 8 | 855 | 104 | |
| BC012604.1 | - | 10 - 11 | 1072 | 4281547 | 104 | |
| Y11651.1 | - | 9 - 10 | 1071 | 104 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000900.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01014192.hg.1 | TC01000900.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014192.hg.1 | ENST00000260563.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014192.hg.1 | ENST00000370128.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014192.hg.1 | NM_001130841.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014192.hg.1 | NM_003729.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | ENST00000260563.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | ENST00000260563.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | ENST00000260563.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | ENST00000260563.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | ENST00000260563.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | ENST00000260563.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | ENST00000260563.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | ENST00000260563.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | ENST00000260563.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | ENST00000260563.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | ENST00000260563.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | ENST00000260563.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | ENST00000260563.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | ENST00000260563.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | ENST00000260563.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | ENST00000370126.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | ENST00000370126.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | ENST00000370126.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | ENST00000370126.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | ENST00000370128.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | ENST00000370128.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | ENST00000370128.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | ENST00000370128.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | ENST00000370128.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | ENST00000370128.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | ENST00000370128.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | ENST00000370128.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | ENST00000370128.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | ENST00000370128.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | ENST00000370128.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | ENST00000370128.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | ENST00000370128.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | ENST00000370128.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | ENST00000370128.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | ENST00000370128.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | ENST00000483474.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | ENST00000483474.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | ENST00000483474.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | ENST00000483474.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | ENST00000483474.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | ENST00000483474.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | ENST00000483474.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | ENST00000483474.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | ENST00000498617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | ENST00000498617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | ENST00000498617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | ENST00000498617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | ENST00000498617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | ENST00000498617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | ENST00000498617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | ENST00000498617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | ENST00000498617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | NM_001130841.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | NM_001130841.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | NM_001130841.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | NM_001130841.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | NM_001130841.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | NM_001130841.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | NM_001130841.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | NM_001130841.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | NM_001130841.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | NM_001130841.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | NM_001130841.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | NM_001130841.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | NM_001130841.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | NM_001130841.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | NM_001130841.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | NM_001130841.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | NM_001130841.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | NM_001130841.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | NM_003729.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | NM_003729.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | NM_003729.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | NM_003729.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | NM_003729.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | NM_003729.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | NM_003729.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | NM_003729.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | NM_003729.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | NM_003729.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | NM_003729.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | NM_003729.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | NM_003729.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | NM_003729.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | NM_003729.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | NM_003729.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009211.hg.1 | NM_003729.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | ENST00000260563.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | ENST00000260563.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | ENST00000260563.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | ENST00000260563.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | ENST00000260563.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | ENST00000260563.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | ENST00000260563.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | ENST00000260563.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | ENST00000260563.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | ENST00000260563.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | ENST00000260563.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | ENST00000260563.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | ENST00000260563.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | ENST00000260563.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | ENST00000260563.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | ENST00000370126.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | ENST00000370126.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | ENST00000370126.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | ENST00000370126.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | ENST00000370128.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | ENST00000370128.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | ENST00000370128.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | ENST00000370128.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | ENST00000370128.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | ENST00000370128.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | ENST00000370128.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | ENST00000370128.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | ENST00000370128.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | ENST00000370128.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | ENST00000370128.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | ENST00000370128.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | ENST00000370128.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | ENST00000370128.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | ENST00000370128.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | ENST00000370128.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | ENST00000483474.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | ENST00000483474.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | ENST00000483474.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | ENST00000483474.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | ENST00000483474.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | ENST00000483474.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | ENST00000483474.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | ENST00000483474.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | ENST00000498617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | ENST00000498617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | ENST00000498617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | ENST00000498617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | ENST00000498617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | ENST00000498617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | ENST00000498617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | ENST00000498617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | ENST00000498617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | NM_001130841.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | NM_001130841.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | NM_001130841.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | NM_001130841.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | NM_001130841.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | NM_001130841.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | NM_001130841.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | NM_001130841.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | NM_001130841.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | NM_001130841.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | NM_001130841.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | NM_001130841.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | NM_001130841.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | NM_001130841.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | NM_001130841.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | NM_001130841.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | NM_001130841.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | NM_001130841.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | NM_003729.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | NM_003729.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | NM_003729.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | NM_003729.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | NM_003729.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | NM_003729.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | NM_003729.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | NM_003729.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | NM_003729.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | NM_003729.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | NM_003729.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | NM_003729.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | NM_003729.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | NM_003729.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | NM_003729.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | NM_003729.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009211.hg.1 | NM_003729.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100165614.hg.1 | ENST00000260563.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100165614.hg.1 | ENST00000370128.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100165614.hg.1 | NM_001130841.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100165614.hg.1 | NM_003729.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100165614.hg.1 | ENST00000260563.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100165614.hg.1 | ENST00000370128.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100165614.hg.1 | NM_001130841.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100165614.hg.1 | NM_003729.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100165614.hg.1 | TC0100009211.hg.1 | Clariom D TC |

RTCA
Genomic Map