Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012319.hg.1 |
ENST00000518732.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012319.hg.1 |
ENST00000518732.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012319.hg.1 |
ENST00000518732.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012319.hg.1 |
ENST00000518732.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012319.hg.1 |
ENST00000518732.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012319.hg.1 |
ENST00000518732.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012319.hg.1 |
ENST00000518732.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012319.hg.1 |
NR_105006.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012319.hg.1 |
NR_105006.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012319.hg.1 |
NR_105006.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012319.hg.1 |
NR_105006.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012319.hg.1 |
NR_105006.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012319.hg.1 |
NR_105006.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012319.hg.1 |
NR_105006.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800185644.hg.1 |
ENST00000518732.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800185644.hg.1 |
NR_105006.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800185644.hg.1 |
ENST00000518732.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800185644.hg.1 |
NR_105006.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800185644.hg.1 |
TC0800012319.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800185643.hg.1 |
ENST00000518732.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800185643.hg.1 |
NR_105006.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800185643.hg.1 |
ENST00000518732.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800185643.hg.1 |
NR_105006.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800185643.hg.1 |
TC0800012319.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800012319.hg.1 |
ENST00000518732.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012319.hg.1 |
ENST00000518732.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012319.hg.1 |
ENST00000518732.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012319.hg.1 |
ENST00000518732.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012319.hg.1 |
ENST00000518732.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012319.hg.1 |
ENST00000518732.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012319.hg.1 |
ENST00000518732.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012319.hg.1 |
NR_105006.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012319.hg.1 |
NR_105006.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012319.hg.1 |
NR_105006.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012319.hg.1 |
NR_105006.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012319.hg.1 |
NR_105006.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012319.hg.1 |
NR_105006.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012319.hg.1 |
NR_105006.1 |
Refseq |