Assay Details
| Gene Symbol: MUC19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK093065.1 | - | 29 - 30 | 1524 | 148 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | leucine rich repeat kinase 2 | LRRK2 |
AURA17, DARDARIN, PARK8, RIPK7, ROCO2 |
-194.2kb | Sense | Chr. 12 - 40224895 - 40369285 on NCBI Build GRCh38 |
| Downstream | contactin 1 | CNTN1 |
F3, GP135, MYPCN |
126.9kb | Sense | Chr. 12 - 40692439 - 41072412 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

MUC19
Genomic Map