Assay Details
| Gene Symbol: SNF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001317192.1 | NP_001304121.1 | 5 - 6 | 528 | 96 | |
| NM_001317193.1 | NP_001304122.1 | 4 - 5 | 477 | 96 | ||
| NM_001317194.1 | NP_001304123.1 | 7 - 8 | 618 | 96 | ||
| NM_007241.3 | NP_009172.2 | 5 - 6 | 528 | 96 | ||
| XM_017024075.1 | XP_016879564.1 | 6 - 7 | 549 | 96 | ||
| GenBank mRNA | AF156102.1 | - | 5 - 6 | 477 | 96 | |
| BC008976.1 | - | 5 - 6 | 478 | 3452153 | 96 | |
| BC038830.1 | - | 5 - 6 | 502 | 3900806 | 96 | |
| BE818533.1 | - | 4 - 5 | 331 | 96 | ||
| BP342490.1 | - | 5 - 6 | 528 | 96 | ||
More Information
Set Membership: |
|

SNF8
Genomic Map