Assay Details
| Gene Symbol: SNF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001317193.1 | NP_001304122.1 | 5 - 6 | 623 | 81 | |
| NM_001317194.1 | NP_001304123.1 | 8 - 9 | 764 | 81 | ||
| NM_007241.3 | NP_009172.2 | 6 - 7 | 674 | 81 | ||
| NR_133679.1 | 6 - 7 | 713 | 81 | |||
| XM_017024075.1 | XP_016879564.1 | 7 - 8 | 695 | 81 | ||
| GenBank mRNA | AF156102.1 | - | 6 - 7 | 623 | 81 | |
| BC008976.1 | - | 6 - 7 | 624 | 3452153 | 81 | |
| BE394098.1 | - | 8 - 9 | 80 | |||
More Information
Set Membership: |
|

SNF8
Genomic Map