Assay Details
| Gene Symbol: PRPF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244926.1 | NP_001231855.1 | 5 - 6 | 677 | 74 | |
| NM_001322266.1 | NP_001309195.1 | 5 - 6 | 677 | 74 | ||
| NM_001322267.1 | NP_001309196.1 | 5 - 6 | 680 | 74 | ||
| NM_004697.4 | NP_004688.2 | 5 - 6 | 680 | 74 | ||
| NR_136265.1 | 5 - 6 | 680 | 74 | |||
| NR_136266.1 | 5 - 6 | 677 | 74 | |||
| GenBank mRNA | AB209797.1 | - | 5 - 6 | 625 | 74 | |
| AF016369.1 | - | 5 - 6 | 630 | 74 | ||
| AK225056.1 | - | 5 - 6 | 653 | 74 | ||
| AK313131.1 | - | 5 - 6 | 622 | 74 | ||
| AY572489.1 | - | 5 - 6 | 567 | 74 | ||
| BC001588.2 | - | 5 - 6 | 620 | 2988225 | 74 | |
| BC007424.2 | - | 5 - 6 | 630 | 3029831 | 74 | |
| CR457105.1 | - | 5 - 6 | 567 | 74 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09000573.hg
Additional Information:
This assay is in the 5'UTR region of the NM_001322266.1,NM_001322267.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR09006370.hg.1 | TC09000573.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006370.hg.1 | ENST00000374198.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006370.hg.1 | ENST00000374199.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006370.hg.1 | NM_001244926.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006370.hg.1 | NM_004697.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006369.hg.1 | TC09000573.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006369.hg.1 | ENST00000374198.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006369.hg.1 | ENST00000374199.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006369.hg.1 | NM_001244926.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006369.hg.1 | NM_004697.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | ENST00000374198.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | ENST00000374198.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | ENST00000374198.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | ENST00000374198.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | ENST00000374198.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | ENST00000374198.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | ENST00000374198.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | ENST00000374198.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | ENST00000374198.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | ENST00000374198.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | ENST00000374198.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | ENST00000374198.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | ENST00000374198.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | ENST00000374198.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | ENST00000374198.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | ENST00000374198.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | ENST00000374198.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | ENST00000374198.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | ENST00000374198.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | ENST00000374198.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | ENST00000374199.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | ENST00000374199.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | ENST00000374199.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | ENST00000374199.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | ENST00000374199.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | ENST00000374199.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | ENST00000374199.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | ENST00000374199.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | ENST00000374199.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | ENST00000374199.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | ENST00000374199.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | ENST00000374199.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | ENST00000374199.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | ENST00000374199.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | ENST00000374199.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | ENST00000374199.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | ENST00000374199.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | ENST00000374199.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | ENST00000374199.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | ENST00000374199.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | ENST00000374199.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | ENST00000374199.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | ENST00000488937.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | ENST00000488937.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | ENST00000488937.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | ENST00000488937.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | NM_001244926.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | NM_001244926.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | NM_001244926.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | NM_001244926.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | NM_001244926.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | NM_001244926.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | NM_001244926.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | NM_001244926.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | NM_001244926.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | NM_001244926.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | NM_001244926.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | NM_001244926.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | NM_001244926.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | NM_001244926.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | NM_001244926.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | NM_001244926.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | NM_001244926.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | NM_001244926.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | NM_001244926.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | NM_001244926.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | NM_001244926.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | NM_004697.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | NM_004697.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | NM_004697.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | NM_004697.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | NM_004697.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | NM_004697.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | NM_004697.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | NM_004697.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | NM_004697.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | NM_004697.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | NM_004697.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | NM_004697.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | NM_004697.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | NM_004697.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | NM_004697.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | NM_004697.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | NM_004697.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | NM_004697.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | NM_004697.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | NM_004697.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008484.hg.1 | NM_004697.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900156649.hg.1 | ENST00000374198.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900156649.hg.1 | ENST00000374199.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900156649.hg.1 | NM_001244926.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900156649.hg.1 | NM_004697.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900156649.hg.1 | ENST00000374198.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900156649.hg.1 | ENST00000374199.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900156649.hg.1 | NM_001244926.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900156649.hg.1 | NM_004697.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900156649.hg.1 | TC0900008484.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | ENST00000374198.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | ENST00000374198.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | ENST00000374198.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | ENST00000374198.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | ENST00000374198.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | ENST00000374198.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | ENST00000374198.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | ENST00000374198.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | ENST00000374198.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | ENST00000374198.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | ENST00000374198.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | ENST00000374198.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | ENST00000374198.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | ENST00000374198.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | ENST00000374198.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | ENST00000374198.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | ENST00000374198.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | ENST00000374198.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | ENST00000374198.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | ENST00000374198.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | ENST00000374199.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | ENST00000374199.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | ENST00000374199.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | ENST00000374199.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | ENST00000374199.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | ENST00000374199.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | ENST00000374199.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | ENST00000374199.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | ENST00000374199.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | ENST00000374199.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | ENST00000374199.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | ENST00000374199.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | ENST00000374199.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | ENST00000374199.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | ENST00000374199.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | ENST00000374199.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | ENST00000374199.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | ENST00000374199.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | ENST00000374199.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | ENST00000374199.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | ENST00000374199.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | ENST00000374199.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | ENST00000488937.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | ENST00000488937.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | ENST00000488937.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | ENST00000488937.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | NM_001244926.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | NM_001244926.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | NM_001244926.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | NM_001244926.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | NM_001244926.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | NM_001244926.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | NM_001244926.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | NM_001244926.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | NM_001244926.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | NM_001244926.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | NM_001244926.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | NM_001244926.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | NM_001244926.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | NM_001244926.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | NM_001244926.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | NM_001244926.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | NM_001244926.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | NM_001244926.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | NM_001244926.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | NM_001244926.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | NM_001244926.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | NM_004697.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | NM_004697.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | NM_004697.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | NM_004697.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | NM_004697.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | NM_004697.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | NM_004697.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | NM_004697.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | NM_004697.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | NM_004697.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | NM_004697.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | NM_004697.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | NM_004697.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | NM_004697.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | NM_004697.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | NM_004697.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | NM_004697.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | NM_004697.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | NM_004697.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | NM_004697.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008484.hg.1 | NM_004697.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900156650.hg.1 | ENST00000374198.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900156650.hg.1 | ENST00000374199.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900156650.hg.1 | NM_001244926.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900156650.hg.1 | NM_004697.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900156650.hg.1 | ENST00000374198.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900156650.hg.1 | ENST00000374199.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900156650.hg.1 | NM_001244926.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900156650.hg.1 | NM_004697.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900156650.hg.1 | TC0900008484.hg.1 | Clariom D TC |

PRPF4
Genomic Map