| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05012738.hg.1 |
TC05000872.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05012738.hg.1 |
ENST00000231198.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05012738.hg.1 |
ENST00000448153.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05012738.hg.1 |
NM_017872.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
ENST00000231198.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
ENST00000231198.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
ENST00000231198.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
ENST00000231198.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
ENST00000231198.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
ENST00000231198.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
ENST00000231198.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
ENST00000231198.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
ENST00000231198.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
ENST00000231198.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
ENST00000231198.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
ENST00000231198.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
ENST00000231198.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
ENST00000521655.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
ENST00000521655.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
ENST00000521655.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
ENST00000521655.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
ENST00000521655.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
ENST00000521655.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
ENST00000521655.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
ENST00000523575.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
ENST00000523575.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
ENST00000523575.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
ENST00000523575.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
ENST00000523575.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
NM_017872.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
NM_017872.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
NM_017872.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
NM_017872.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
NM_017872.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
NM_017872.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
NM_017872.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
NM_017872.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
NM_017872.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
NM_017872.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
NM_017872.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
ENST00000231198.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
ENST00000231198.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
ENST00000231198.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
ENST00000231198.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
ENST00000231198.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
ENST00000231198.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
ENST00000231198.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
ENST00000231198.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
ENST00000231198.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
ENST00000231198.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
ENST00000231198.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
ENST00000231198.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
ENST00000231198.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
ENST00000521655.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
ENST00000521655.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
ENST00000521655.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
ENST00000521655.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
ENST00000521655.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
ENST00000521655.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
ENST00000521655.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
ENST00000523575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
ENST00000523575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
ENST00000523575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
ENST00000523575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
ENST00000523575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
NM_017872.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
NM_017872.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
NM_017872.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
NM_017872.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
NM_017872.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
NM_017872.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
NM_017872.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
NM_017872.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
NM_017872.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
NM_017872.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
NM_017872.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500164604.hg.1 |
ENST00000231198.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500164604.hg.1 |
ENST00000521655.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500164604.hg.1 |
ENST00000523575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500164604.hg.1 |
NM_017872.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500164604.hg.1 |
ENST00000231198.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500164604.hg.1 |
ENST00000521655.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500164604.hg.1 |
ENST00000523575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500164604.hg.1 |
NM_017872.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500164604.hg.1 |
TC0500009223.hg.1 |
Clariom D TC |