Assay Details
| Gene Symbol: BNIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205.2 | NP_001196.2 | 2 - 3 | 279 | 114 | |
| NM_013978.2 | NP_053581.2 | 2 - 3 | 279 | 114 | ||
| GenBank mRNA | AF083956.1 | - | 2 - 3 | 175 | 114 | |
| BC010959.1 | - | 2 - 3 | 204 | 4052817 | 114 | |
| U15172.1 | - | 2 - 3 | 185 | 114 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05000959.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05013957.hg.1 | TC05000959.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05013957.hg.1 | ENST00000231668.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05013957.hg.1 | ENST00000351486.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05013957.hg.1 | ENST00000352523.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05013957.hg.1 | ENST00000393770.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05013957.hg.1 | NM_001205.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05013957.hg.1 | NM_013978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05013957.hg.1 | NM_013979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05013957.hg.1 | NM_013980.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000231668.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000231668.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000231668.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000231668.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000231668.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000231668.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000231668.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000231668.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000231668.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000231668.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000231668.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000351486.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000351486.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000351486.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000351486.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000351486.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000351486.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000351486.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000351486.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000351486.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000351486.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000352523.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000352523.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000352523.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000352523.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000352523.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000352523.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000352523.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000352523.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000352523.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000393770.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000393770.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000393770.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000393770.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000393770.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000393770.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000393770.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000519459.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | ENST00000519459.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_001205.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_001205.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_001205.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_001205.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_001205.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_001205.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_001205.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_001205.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_001205.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_001205.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_001205.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013979.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013980.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013980.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013980.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013980.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013980.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013980.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013980.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013980.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013980.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013980.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009490.hg.1 | NM_013980.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500166421.hg.1 | ENST00000231668.13 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500166421.hg.1 | ENST00000351486.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500166421.hg.1 | ENST00000352523.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500166421.hg.1 | ENST00000393770.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500166421.hg.1 | NM_001205.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500166421.hg.1 | NM_013978.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500166421.hg.1 | NM_013979.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500166421.hg.1 | NM_013980.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500166421.hg.1 | ENST00000231668.13 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500166421.hg.1 | ENST00000351486.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500166421.hg.1 | ENST00000352523.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500166421.hg.1 | ENST00000393770.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500166421.hg.1 | NM_001205.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500166421.hg.1 | NM_013978.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500166421.hg.1 | NM_013979.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500166421.hg.1 | NM_013980.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500166421.hg.1 | TC0500009490.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000231668.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000231668.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000231668.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000231668.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000231668.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000231668.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000231668.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000231668.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000231668.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000231668.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000231668.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000351486.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000351486.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000351486.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000351486.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000351486.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000351486.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000351486.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000351486.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000351486.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000351486.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000352523.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000352523.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000352523.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000352523.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000352523.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000352523.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000352523.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000352523.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000352523.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000393770.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000393770.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000393770.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000393770.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000393770.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000393770.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000393770.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000519459.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | ENST00000519459.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_001205.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_001205.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_001205.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_001205.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_001205.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_001205.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_001205.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_001205.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_001205.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_001205.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_001205.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013978.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013978.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013978.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013978.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013978.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013978.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013978.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013978.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013978.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013978.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013979.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013980.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013980.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013980.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013980.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013980.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013980.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013980.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013980.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013980.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013980.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009490.hg.1 | NM_013980.2 | Refseq |

BNIP1
Genomic Map