Assay Details
| Gene Symbol: PAX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000438.5 | NP_000429.2 | 3 - 4 | 826 | 55 | |
| NM_001127366.2 | NP_001120838.1 | 3 - 4 | 823 | 55 | ||
| NM_013942.4 | NP_039230.1 | 3 - 4 | 826 | 55 | ||
| NM_181457.3 | NP_852122.1 | 3 - 4 | 826 | 55 | ||
| NM_181458.3 | NP_852123.1 | 3 - 4 | 826 | 55 | ||
| NM_181459.3 | NP_852124.1 | 3 - 4 | 826 | 55 | ||
| NM_181460.3 | NP_852125.1 | 3 - 4 | 826 | 55 | ||
| NM_181461.3 | NP_852126.1 | 3 - 4 | 826 | 55 | ||
| GenBank mRNA | AI399816.1 | - | 2 - 3 | 264 | 55 | |
| AK291278.1 | - | 3 - 4 | 826 | 55 | ||
| AY251279.1 | - | 3 - 4 | 496 | 55 | ||
| AY251280.1 | - | 3 - 4 | 496 | 55 | ||
| BC063547.1 | - | 3 - 4 | 695 | 55 | ||
| BC101299.3 | - | 3 - 4 | 722 | 40024655 | 55 | |
| BC101300.2 | - | 3 - 4 | 722 | 40024656 | 55 | |
| BC101301.2 | - | 3 - 4 | 725 | 40024658 | 55 | |
| BC101302.2 | - | 3 - 4 | 722 | 40024661 | 55 | |
| BC114363.2 | - | 3 - 4 | 722 | 40024665 | 55 | |
| S69369.1 | - | 3 - 4 | 576 | 55 | ||
| S69370.1 | - | 3 - 4 | 576 | 55 | ||
More Information
Set Membership: |
|

PAX3
Genomic Map