Assay Details
| Gene Symbol: PAX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127366.2 | NP_001120838.1 | 4 - 5 | 962 | 103 | |
| NM_181457.3 | NP_852122.1 | 4 - 5 | 965 | 103 | ||
| NM_181458.3 | NP_852123.1 | 4 - 5 | 965 | 103 | ||
| NM_181459.3 | NP_852124.1 | 4 - 5 | 965 | 103 | ||
| NM_181460.3 | NP_852125.1 | 4 - 5 | 965 | 103 | ||
| NM_181461.3 | NP_852126.1 | 4 - 5 | 965 | 103 | ||
| GenBank mRNA | AK291278.1 | - | 4 - 5 | 965 | 103 | |
| AY251279.1 | - | 4 - 5 | 635 | 103 | ||
| AY251280.1 | - | 4 - 5 | 635 | 103 | ||
| BC101299.3 | - | 4 - 5 | 861 | 40024655 | 103 | |
| BC101300.2 | - | 4 - 5 | 861 | 40024656 | 103 | |
| BC101301.2 | - | 4 - 5 | 864 | 40024658 | 103 | |
| BC101302.2 | - | 4 - 5 | 861 | 40024661 | 103 | |
| BC114363.2 | - | 4 - 5 | 861 | 40024665 | 103 | |
| L07483.1 | - | 1 - 2 | 41 | 103 | ||
| U02309.1 | - | 1 - 2 | 140 | 103 | ||
More Information
Set Membership: |
|

PAX3
Genomic Map