Assay Details
| Gene Symbol: IL32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001308078.1 | NP_001295007.1 | 3 - 4 | 319 | 119 | |
| GenBank mRNA | BC009401.2 | - | 3 - 4 | 456 | 4300407 | 119 |
| BF568456.1 | - | 3 - 4 | 457 | 119 | ||
| M59807.1 | - | 3 - 4 | 304 | 119 | ||
More Information
Set Membership: |
|

IL32
Genomic Map