Assay Details
| Gene Symbol: P4HA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142598.1 | NP_001136070.1 | 2 - 3 | 213 | 70 | |
| NM_001142599.1 | NP_001136071.1 | 2 - 3 | 213 | 70 | ||
| XM_005272117.4 | XP_005272174.1 | 2 - 3 | 235 | 70 | ||
| XM_005272118.4 | XP_005272175.1 | 2 - 3 | 364 | 70 | ||
| XM_006714729.3 | XP_006714792.1 | 2 - 3 | 235 | 70 | ||
| GenBank mRNA | AY358970.1 | - | 1 - 2 | 151 | 70 | |
| BC017062.1 | - | 1 - 2 | 179 | 3847723 | 70 | |
| BI758477.1 | - | 2 - 3 | 164 | 70 | ||
| DA544450.1 | - | 2 - 3 | 213 | 70 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AY358970.1, BC017062.1, NM_001142598.1, NM_001142599.1, XM_005272117.4, XM_005272118.4, XM_006714729.3 transcripts.
Set Membership: |
|

P4HA2
Genomic Map