Assay Details
| Gene Symbol: NRG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013981.3 | NP_053584.1 | 5 - 6 | 1399 | 64 | |
| XM_005268533.3 | XP_005268590.1 | 5 - 6 | 477 | 64 | ||
| XM_017010066.1 | XP_016865555.1 | 6 - 7 | 1841 | 64 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005268533.3 transcripts.
Set Membership: |
|

NRG2
Genomic Map