| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05025366.hg.1 |
TC05001851.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05025366.hg.1 |
ENST00000289409.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05025366.hg.1 |
ENST00000289422.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05025366.hg.1 |
ENST00000340391.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05025366.hg.1 |
ENST00000358522.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05025366.hg.1 |
ENST00000361474.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05025366.hg.1 |
ENST00000394770.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05025366.hg.1 |
ENST00000446269.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05025366.hg.1 |
ENST00000541337.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05025366.hg.1 |
ENST00000544729.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05025366.hg.1 |
ENST00000545385.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05025366.hg.1 |
NM_001184935.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05025366.hg.1 |
NM_004883.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05025366.hg.1 |
NM_013981.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05025366.hg.1 |
NM_013982.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05025366.hg.1 |
NM_013983.2 |
Refseq |