Assay Details
| Gene Symbol: NRG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001184935.1 | NP_001171864.1 | 1 | 772 | 104 | |
| NM_004883.2 | NP_004874.1 | 1 | 772 | 104 | ||
| NM_013981.3 | NP_053584.1 | 1 | 772 | 104 | ||
| NM_013982.2 | NP_053585.1 | 1 | 772 | 104 | ||
| NM_013983.2 | NP_053586.1 | 1 | 772 | 104 | ||
| XM_006714810.3 | XP_006714873.1 | 1 | 1138 | 104 | ||
| XM_011537713.2 | XP_011536015.1 | 1 | 1148 | 104 | ||
| XM_017010066.1 | XP_016865555.1 | 1 | 1137 | 104 | ||
| GenBank mRNA | AB005060.1 | - | 1 | 767 | 104 | |
| AK124504.1 | - | 2 | 701 | 104 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05001851.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
signal transduction
regulation of phosphatidylinositol 3-kinase signaling
peptidyl-tyrosine phosphorylation
intracellular signal transduction
ERBB2 signaling pathway
positive regulation of GTPase activity
phosphatidylinositol phosphorylation
phosphatidylinositol-mediated signaling
animal organ development
regulation of cell motility
protein tyrosine kinase activity
Ras guanyl-nucleotide exchange factor activity
receptor binding
growth factor activity
phosphatidylinositol-4,5-bisphosphate 3-kinase activity
MAPK cascade
signal transduction
regulation of phosphatidylinositol 3-kinase signaling
peptidyl-tyrosine phosphorylation
intracellular signal transduction
ERBB2 signaling pathway
positive regulation of GTPase activity
phosphatidylinositol phosphorylation
phosphatidylinositol-mediated signaling
animal organ development
regulation of cell motility
protein tyrosine kinase activity
Ras guanyl-nucleotide exchange factor activity
receptor binding
growth factor activity
phosphatidylinositol-4,5-bisphosphate 3-kinase activity
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05025386.hg.1 | TC05001851.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025386.hg.1 | ENST00000289422.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025386.hg.1 | ENST00000361474.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025386.hg.1 | ENST00000394770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025386.hg.1 | ENST00000446269.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025386.hg.1 | ENST00000541337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025386.hg.1 | ENST00000544729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025386.hg.1 | ENST00000545385.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025386.hg.1 | NM_001184935.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025386.hg.1 | NM_004883.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025386.hg.1 | NM_013981.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025386.hg.1 | NM_013982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025386.hg.1 | NM_013983.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025387.hg.1 | TC05001851.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025387.hg.1 | ENST00000361474.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025387.hg.1 | ENST00000394770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025387.hg.1 | ENST00000446269.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025387.hg.1 | ENST00000541337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025387.hg.1 | ENST00000544729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025387.hg.1 | ENST00000545385.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025387.hg.1 | NM_001184935.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025387.hg.1 | NM_004883.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025387.hg.1 | NM_013981.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025387.hg.1 | NM_013982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025387.hg.1 | NM_013983.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025384.hg.1 | TC05001851.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025384.hg.1 | ENST00000289409.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025384.hg.1 | ENST00000289422.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025384.hg.1 | ENST00000358522.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025384.hg.1 | ENST00000361474.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025384.hg.1 | ENST00000394770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025384.hg.1 | ENST00000446269.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025384.hg.1 | ENST00000541337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025384.hg.1 | ENST00000545385.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025384.hg.1 | NM_001184935.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025384.hg.1 | NM_004883.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025384.hg.1 | NM_013981.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025384.hg.1 | NM_013982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025384.hg.1 | NM_013983.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025383.hg.1 | TC05001851.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025383.hg.1 | ENST00000289409.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025383.hg.1 | ENST00000289422.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025383.hg.1 | ENST00000358522.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025383.hg.1 | ENST00000361474.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025383.hg.1 | ENST00000394770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025383.hg.1 | ENST00000446269.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025383.hg.1 | ENST00000541337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025383.hg.1 | ENST00000544729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025383.hg.1 | ENST00000545385.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025383.hg.1 | NM_001184935.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025383.hg.1 | NM_004883.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025383.hg.1 | NM_013981.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025383.hg.1 | NM_013982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025383.hg.1 | NM_013983.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025385.hg.1 | TC05001851.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025385.hg.1 | ENST00000289409.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025385.hg.1 | ENST00000289422.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025385.hg.1 | ENST00000358522.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025385.hg.1 | ENST00000361474.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025385.hg.1 | ENST00000394770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025385.hg.1 | ENST00000446269.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025385.hg.1 | ENST00000541337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025385.hg.1 | ENST00000544729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025385.hg.1 | ENST00000545385.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025385.hg.1 | NM_001184935.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025385.hg.1 | NM_004883.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025385.hg.1 | NM_013981.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025385.hg.1 | NM_013982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05025385.hg.1 | NM_013983.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500184869.hg.1 | ENST00000361474.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500184869.hg.1 | ENST00000541337.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500184869.hg.1 | NM_001184935.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500184869.hg.1 | NM_004883.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500184869.hg.1 | NM_013981.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500184869.hg.1 | NM_013982.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500184869.hg.1 | NM_013983.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500184869.hg.1 | ENST00000361474.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500184869.hg.1 | ENST00000541337.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500184869.hg.1 | NM_001184935.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500184869.hg.1 | NM_004883.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500184869.hg.1 | NM_013981.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500184869.hg.1 | NM_013982.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500184869.hg.1 | NM_013983.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500184869.hg.1 | TC0500012223.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500184865.hg.1 | ENST00000289409.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500184865.hg.1 | ENST00000289422.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500184865.hg.1 | ENST00000358522.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500184865.hg.1 | ENST00000361474.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500184865.hg.1 | ENST00000378238.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500184865.hg.1 | ENST00000541337.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500184865.hg.1 | NM_001184935.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500184865.hg.1 | NM_004883.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500184865.hg.1 | NM_013981.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500184865.hg.1 | NM_013982.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500184865.hg.1 | NM_013983.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500184865.hg.1 | ENST00000289409.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500184865.hg.1 | ENST00000289422.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500184865.hg.1 | ENST00000358522.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500184865.hg.1 | ENST00000361474.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500184865.hg.1 | ENST00000378238.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500184865.hg.1 | ENST00000541337.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500184865.hg.1 | NM_001184935.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500184865.hg.1 | NM_004883.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500184865.hg.1 | NM_013981.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500184865.hg.1 | NM_013982.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500184865.hg.1 | NM_013983.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500184865.hg.1 | TC0500012223.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500184867.hg.1 | ENST00000289422.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500184867.hg.1 | ENST00000361474.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500184867.hg.1 | ENST00000541337.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500184867.hg.1 | NM_001184935.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500184867.hg.1 | NM_004883.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500184867.hg.1 | NM_013981.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500184867.hg.1 | NM_013982.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500184867.hg.1 | NM_013983.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500184867.hg.1 | ENST00000289422.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500184867.hg.1 | ENST00000361474.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500184867.hg.1 | ENST00000541337.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500184867.hg.1 | NM_001184935.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500184867.hg.1 | NM_004883.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500184867.hg.1 | NM_013981.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500184867.hg.1 | NM_013982.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500184867.hg.1 | NM_013983.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500184867.hg.1 | TC0500012223.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500184870.hg.1 | ENST00000361474.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500184870.hg.1 | ENST00000541337.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500184870.hg.1 | NM_001184935.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500184870.hg.1 | NM_004883.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500184870.hg.1 | NM_013981.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500184870.hg.1 | NM_013982.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500184870.hg.1 | NM_013983.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500184870.hg.1 | ENST00000361474.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500184870.hg.1 | ENST00000541337.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500184870.hg.1 | NM_001184935.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500184870.hg.1 | NM_004883.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500184870.hg.1 | NM_013981.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500184870.hg.1 | NM_013982.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500184870.hg.1 | NM_013983.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500184870.hg.1 | TC0500012223.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500184866.hg.1 | ENST00000289409.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500184866.hg.1 | ENST00000289422.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500184866.hg.1 | ENST00000358522.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500184866.hg.1 | ENST00000361474.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500184866.hg.1 | ENST00000378238.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500184866.hg.1 | ENST00000541337.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500184866.hg.1 | NM_001184935.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500184866.hg.1 | NM_004883.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500184866.hg.1 | NM_013981.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500184866.hg.1 | NM_013982.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500184866.hg.1 | NM_013983.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500184866.hg.1 | ENST00000289409.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500184866.hg.1 | ENST00000289422.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500184866.hg.1 | ENST00000358522.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500184866.hg.1 | ENST00000361474.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500184866.hg.1 | ENST00000378238.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500184866.hg.1 | ENST00000541337.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500184866.hg.1 | NM_001184935.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500184866.hg.1 | NM_004883.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500184866.hg.1 | NM_013981.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500184866.hg.1 | NM_013982.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500184866.hg.1 | NM_013983.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500184866.hg.1 | TC0500012223.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 170215 | - | - |

NRG2
Genomic Map