Assay Details
| Gene Symbol: DNASE1L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011522399.1 | XP_011520701.1 | 1 | 196 | 76 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BE257515.1 | - | 1 | Chr. 16 - 2236304 - 2236894 [-] on Build GRCh38 | 76 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | E4F transcription factor 1 | E4F1 | E4F | -510.0 | Sense | Chr. 16 - 2223488 - 2235742 on NCBI Build GRCh38 |
| Downstream | enoyl-CoA delta isomerase 1 | ECI1 | DCI | 3.3kb | Anti-sense | Chr. 16 - 2239872 - 2251601 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000320700.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000320700.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000320700.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000320700.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000320700.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000320700.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000320700.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000320700.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000320700.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000320700.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000320700.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000382437.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000382437.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000382437.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000382437.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000382437.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000382437.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000382437.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000382437.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000382437.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000382437.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000564065.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000564065.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000564065.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000564065.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000564065.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000564065.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000564065.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000564065.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000564065.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000564065.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000564065.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000564065.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000564065.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000564065.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000567494.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000567494.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000567494.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000567494.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000567494.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000567494.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000567494.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000567494.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000567494.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000567494.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000567494.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000569184.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000569184.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000569184.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000569184.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000569184.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000569184.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000569184.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000613572.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000613572.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000613572.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000613572.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000613572.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000613572.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000613572.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000613572.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000613572.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | ENST00000613572.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | NM_001301680.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | NM_001301680.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | NM_001301680.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | NM_001301680.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | NM_001301680.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | NM_001301680.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | NM_001301680.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | NM_001301680.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | NM_001301680.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | NM_001301680.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | NM_001301680.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | NM_001374.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | NM_001374.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | NM_001374.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | NM_001374.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | NM_001374.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | NM_001374.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | NM_001374.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | NM_001374.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | NM_001374.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | NM_001374.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006601.hg.1 | NM_001374.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000320700.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000320700.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000320700.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000320700.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000320700.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000320700.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000320700.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000320700.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000320700.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000320700.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000320700.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000382437.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000382437.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000382437.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000382437.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000382437.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000382437.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000382437.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000382437.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000382437.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000382437.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000564065.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000564065.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000564065.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000564065.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000564065.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000564065.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000564065.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000564065.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000564065.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000564065.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000564065.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000564065.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000564065.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000564065.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000567494.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000567494.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000567494.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000567494.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000567494.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000567494.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000567494.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000567494.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000567494.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000567494.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000567494.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000569184.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000569184.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000569184.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000569184.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000569184.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000569184.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000569184.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000613572.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000613572.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000613572.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000613572.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000613572.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000613572.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000613572.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000613572.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000613572.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | ENST00000613572.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | NM_001301680.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | NM_001301680.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | NM_001301680.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | NM_001301680.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | NM_001301680.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | NM_001301680.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | NM_001301680.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | NM_001301680.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | NM_001301680.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | NM_001301680.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | NM_001301680.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | NM_001374.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | NM_001374.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | NM_001374.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | NM_001374.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | NM_001374.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | NM_001374.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | NM_001374.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | NM_001374.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | NM_001374.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | NM_001374.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006601.hg.1 | NM_001374.2 | Refseq |

DNASE1L2
Genomic Map